Merck
  • Design, synthesis and biological evaluation of small molecules as potent glucosidase inhibitors.
Design, synthesis and biological evaluation of small molecules as potent glucosidase inhibitors.
European journal of medicinal chemistry (2015-06-19)
Santanu Hati, Sanjay M Madurkar, Chandramohan Bathula, Chiranjeevi Thulluri, Rahul Agarwal, Faiza Amber Siddiqui, Poonam Dangi, Uma Adepally, Ashutosh Singh, Shailja Singh, Subhabrata Sen
ABSTRACT
Herein we have reported design, synthesis and in vitro biological evaluation of a library of bicyclic lactams that led to the discovery of compounds 6 and 7 as a novel class of α-glucosidase inhibitors. They inhibited α-glucosidase (yeast origin) in a mixed type of inhibition with an IC50 of ∼150 nM. Molecular docking studies further substantiated screening results. Interestingly phenotypic screening of this library against the human malaria parasite revealed 7 as a potent antiplasmodial agent.
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide, 97%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Potassium permanganate, meets USP testing specifications
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.5 M in THF
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.0 M in THF
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.0 M in hexanes
Sigma-Aldrich
Phenylselenyl bromide, 98%
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1 M in tert-butyl methyl ether
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 0.5 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1 M in toluene