Merck
  • JARID2 inhibits leukemia cell proliferation by regulating CCND1 expression.
JARID2 inhibits leukemia cell proliferation by regulating CCND1 expression.
International journal of hematology (2015-05-06)
Chang-Liang Su, Tao-Ran Deng, Zhen Shang, Yi Xiao
ABSTRACT
It has recently been shown that JARID2 contributes to the malignant character of solid tumors, such as epithelial-mesenchymal transition in lung and colon cancer cell lines, but its role in leukemia progression is unexplored. In this study, we explored the effect and underlying molecular mechanism of JARID2 on leukemia cell proliferation. Real-time PCR and Western assay were carried out to detect JARID2 and CCND1 expression. Cell number and cell cycle change were detected using hemocytometer and flow cytometry, and a ChIP assay was utilized to investigate JARID2 and H3K27me3 enrichment on the CCND1 promoter. JARID2 is down-regulated in B-chronic lymphocytic leukemia (B-CLL) and acute monocytic leukemia (AMOL), and knockdown of JARID2 promotes leukemia cell proliferation via acceleration of the G1/S transition. Conversely, ectopic expression of JARID2 inhibits these malignant phenotypes. Mechanistic studies show that JARID2 negatively regulates CCND1 expression by increasing H3K27 trimethylation on the CCND1 promoter. Our findings indicate that JARID2 is a negative regulator of leukemia cell proliferation, and functions as potential tumor suppressor in leukemia.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
MISSION® esiRNA, targeting human JARID2