Merck
  • Hydrogen sulfide protects from colitis and restores intestinal microbiota biofilm and mucus production.
Hydrogen sulfide protects from colitis and restores intestinal microbiota biofilm and mucus production.
Inflammatory bowel diseases (2015-03-05)
Jean-Paul Motta, Kyle L Flannigan, Terence A Agbor, Jennifer K Beatty, Rory W Blackler, Matthew L Workentine, Gabriela J Da Silva, Rui Wang, Andre G Buret, John L Wallace
ABSTRACT
Microbiota dysbiosis and impaired barrier function are among the most prominent features of inflammatory bowel disease. In the gastrointestinal tract, hydrogen sulfide (H(2)S) is an important regulator of mucosal homeostasis. We hypothesized that H(2)S promotes resolution of colonic inflammation through actions on microbiota biofilm and the mucus barrier. We used mice genetically deficient for a key enzyme for H(2)S production (cystathionine γ-lyase) and pharmacologically inhibited that enzyme during colitis in wild-type mice. We tested the effects of administering an H(2)S donor (diallyl disulfide) to rodents during hapten-induced colitis. Colonic microbiota biofilm was visualized by fluorescent in situ hybridization, and mucus granules were quantified with periodic acid-alcian blue staining. We exposed human microbiota biofilms and planktonic bacteria to H(2)S donors ex vivo to determine changes in their growth, viability, and biomass. Intestinal microbiota formed linear biofilms in the colon of healthy rodents. During colitis, microbiota biofilms were fragmented and mucus granule production decreased. Endogenous production of H(2)S had beneficial effects on establishment of microbiota biofilms and colonic mucus production. Therapeutic delivery of H(2)S into the colon reduced inflammation, restored the microbiota biofilm, and increased the production of mucus granules. In ex vivo human microbiota, H(2)S not only promoted biofilm formation but also reduced growth of planktonic bacteria. Our results suggest that H(2)S donors could be used therapeutically during colitis, facilitating correction of microbiota biofilm dysbiosis and mucus layer reconstitution.
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
Hydrogen sulfide, ≥99.5%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
Sulfur-32S, 99.9 atom % 32S
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
Allyl disulfide, technical grade, 80%
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
Allyl disulfide, ≥80%, FG
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Menadione, meets USP testing specifications
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
DAPI, for nucleic acid staining
Supelco
Cannabidiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Hydrogen sulfide solution, 0.8 M in THF