Merck
  • DNA-Linked Enzyme-Coupled Assay for Probing Glucosyltransferase Specificity.
DNA-Linked Enzyme-Coupled Assay for Probing Glucosyltransferase Specificity.
ACS synthetic biology (2015-01-27)
David J Sukovich, Cyrus Modavi, Markus de Raad, Robin N Prince, J Christopher Anderson
ABSTRACT
Traditional enzyme characterization methods are low-throughput and therefore limit engineering efforts in synthetic biology and biotechnology. Here, we propose a DNA-linked enzyme-coupled assay (DLEnCA) to monitor enzyme reactions in a high-throughput manner. Throughput is improved by removing the need for protein purification and by limiting the need for liquid chromatography mass spectrometry (LCMS) product detection by linking enzymatic function to DNA modification. We demonstrate the DLEnCA methodology using glucosyltransferases as an illustration. The assay utilizes cell free transcription/translation systems to produce enzymes of interest, while UDP-glucose and T4-β-glucosyltransferase are used to modify DNA, which is detected postreaction using qPCR or a similar means of DNA analysis. OleD and two glucosyltransferases from Arabidopsis were used to verify the assay's generality toward glucosyltransferases. We further show DLEnCA's utility by mapping out the substrate specificity for these enzymes.
Sigma-Aldrich
L-(+)-Arabinose, 99%
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Sigma-Aldrich
4-Methylumbelliferone, ≥98%
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
L-(+)-Arabinose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
7-Hydroxycoumarinyl-4-acetic acid, 97%
Sigma-Aldrich
L-(+)-Arabinose, 98%
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
3-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
3-Hydroxyflavone, ≥98%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ultrapure Acetonitrile