Skip to Content
Merck
All Photos(1)

Documents

335665

Sigma-Aldrich

7-Hydroxycoumarinyl-4-acetic acid

97%

Synonym(s):

Umbelliferone-4-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8O5
CAS Number:
Molecular Weight:
220.18
Beilstein:
204777
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

212 °C (dec.) (lit.)

SMILES string

OC(=O)CC1=CC(=O)Oc2cc(O)ccc12

InChI

1S/C11H8O5/c12-7-1-2-8-6(3-10(13)14)4-11(15)16-9(8)5-7/h1-2,4-5,12H,3H2,(H,13,14)

InChI key

BNHPMQBVNXMPDU-UHFFFAOYSA-N

Application

7-Hydroxycoumarinyl-4-acetic acid was used in the synthesis of novel and specific enzyme substrates for Maroteaux-Lamy (MPS VI) and Morquio A (MPS IVA) diseases. It was also employed as ionic liquid matrice for MALDI-TOF MS of proteins, peptides and carbohydrates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Erland J F Demant et al.
The Biochemical journal, 363(Pt 3), 809-815 (2002-04-20)
The kinetics of the interaction of long-chain fatty acids (referred to as fatty acids) with albumin is critical to understanding the role of albumin in fatty acid transport. In this study we have determined the kinetics of fatty acid dissociation
E J Demant et al.
Analytical biochemistry, 295(1), 38-44 (2001-07-31)
The fluorescence-based long-chain fatty acid probe BSA-HCA (bovine serum albumin labeled with 7-hydroxycoumarin-4-acetic acid) is shown to respond to binding of long-chain acyl-CoA thioesters by quenching of the 450 nm fluorescence emission. As determined by spectrofluorometric titration, binding affinities for
Trisha A Duffey et al.
Bioorganic & medicinal chemistry letters, 20(20), 5994-5996 (2010-09-14)
In continued efforts to develop enzymatic assays for lysosomal storage diseases appropriate for newborn screening laboratories we have synthesized novel and specific enzyme substrates for Maroteaux-Lamy (MPS VI) and Morquio A (MPS IVA) diseases. The sulfated monosaccharide derivatives were found
Yan Chen et al.
Analytical biochemistry, 540-541, 38-44 (2017-11-13)
To establish a high throughput, low cost, and simple nanotechnology-based method for the detection of single nucleotide polymorphism (SNP) loci in type 2 diabetes mellitus (T2DM). Multiplex ligase detection reaction (LDR) amplification was performed using fluorescently labeled magnetic nanosphere-bound upstream
E J Demant
Analytical biochemistry, 267(2), 366-372 (1999-02-26)
A covalent complex between bovine serum albumin and 7-hydroxycoumarin-4-acetic acid (BSA-HCA) shows a strong fluorescence band at lambdamax = 450 nm upon excitation at 375 nm. Quenching of the fluorescence emission accompanies the association of fatty acids (FA) to BSA-HCA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service