Merck
  • Physical and functional interaction of the TPL2 kinase with nucleophosmin.
Physical and functional interaction of the TPL2 kinase with nucleophosmin.
Oncogene (2014-07-08)
D C Kanellis, S Bursac, P N Tsichlis, S Volarevic, A G Eliopoulos
ABSTRACT
Tumor Progression Locus 2 (TPL2) is widely recognized as a cytoplasmic mitogen-activated protein 3 kinase with a prominent role in the regulation of inflammatory and oncogenic signal transduction. Herein we report that TPL2 may also operate in the nucleus as a physical and functional partner of nucleophosmin (NPM/B23), a major nucleolar phosphoprotein with diverse cellular activities linked to malignancy. We demonstrate that TPL2 mediates the phosphorylation of a fraction of NPM at threonine 199, an event required for its proteasomal degradation and maintenance of steady-state NPM levels. Upon exposure to ultraviolet C, Tpl2 is required for the translocation of de-phosphorylated NPM from the nucleolus to the nucleoplasm. NPM is an endogenous inhibitor of HDM2:p53 interaction and knockdown of TPL2 was found to result in reduced binding of NPM to HDM2, with concomitant defects in p53 accumulation following genotoxic or ribosomal stress. These findings expand our understanding of the function of TPL2 as a negative regulator of carcinogenesis by defining a nuclear role for this kinase in the topological sequestration of NPM, linking p53 signaling to the generation of threonine 199-phosphorylated NPM.
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse, clone MAPK-YT, ascites fluid
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Map3k8
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human MAP3K8
Sigma-Aldrich
N-Ethylmaleimide, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Ethylmaleimide, BioXtra, ≥98% (HPLC)