Merck
Cationic Xylene Tag for Increasing Sensitivity in Mass Spectrometry.
Journal of the American Society for Mass Spectrometry (2015-06-28)
Poguang Wang, Qi Zhang, Yuanyuan Yao, Roger W Giese
ABSTRACT
N-(2-(Bromomethyl)benzyl)-N,N-diethylethanaminium bromide, that we designate as CAX-B (cationic xylyl-bromide), is presented as a derivatization reagent for increasing sensitivity in mass spectrometry. Because of its aryl bromomethyl moiety, CAX-B readily labels compounds having an active hydrogen. In part, a CAX-tagged analyte (CAX-analyte) can be very sensitive especially in a tandem mass spectrometer (both ESI and MALDI). This is because of facile formation of an analyte-characteristic first product ion (as a xylyl-based cation) from favorable loss of triethylamine as a neutral from the precursor ion. This loss is enhanced both by resonance stabilization of the xylyl cation, and by anchimeric assistance from the ortho hetero atom of the attached analyte. High intensity of a first product ion opens up the opportunity for a CAX-analyte to be additionally sensitive when it is prone to a secondary neutral loss from the analyte part. For example, we have derivatized and detected 160 amol of thymidine by CAX-tagging/LC-MALDI-TOF/TOF-MS in this way, where the two neutral losses are triethylamine and deoxyribose. Other analytes detected at the amol level as CAX derivatives (as diluted standards) include estradiol and some nucleobases. The tendency for analytes with multiple active hydrogens to label just once with CAX (an advantage) is illustrated by the conversion of bisphenol A to a single product even when excess CAX-B is present. A family of analogous reagents with a variety of reactivity groups is anticipated as a consequence of replacing the bromine atom of CAX-B with various functional groups.
Sigma-Aldrich
o-Xylene-d10, 99 atom % D
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Ethyl acetate
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Bromine, ≥99.99% trace metals basis
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Bisphenol A, 97%
Sigma-Aldrich
p-Xylene, anhydrous, ≥99%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Thymidine, ≥99%