Merck
  • Occurrence and human exposure of parabens and their chlorinated derivatives in swimming pools.
Occurrence and human exposure of parabens and their chlorinated derivatives in swimming pools.
Environmental science and pollution research international (2015-07-15)
Wenhui Li, Yali Shi, Lihong Gao, Jiemin Liu, Yaqi Cai
ABSTRACT
As an emerging group of endocrine-disrupting chemicals, parabens have attracted growing attention due to their potential effects on human health. In the present study, the occurrence and distribution of eight parabens, four chlorinated parabens, and their common hydrolysis product, p-hydroxybenzoic acid (PHBA), were investigated in 39 swimming pools in Beijing, China. Methyl paraben and propyl paraben were the predominant compounds in swimming pools, accounting for 91.2 % of the total parabens. It is noteworthy that octyl paraben, a paraben with longer chain, was firstly detected in this study. There were several factors affecting the levels of parabens among the 39 swimming pools. The concentrations of parabens and chlorinated derivatives detected in indoor pools (144 ng L(-1)) were roughly 20-fold higher than those in outdoor pools (6.78 ng L(-1)). Hotel pools appear to present higher level of target compounds (361 ng L(-1)) than that in health club (228 ng L(-1)), municipal (130 ng L(-1)), school (75.6 ng L(-1)), and community pools (63.0 ng L(-1)). Moreover, the level of these compounds in pools during weekends (174 ng L(-1)) was much higher than that during weekdays (52.3 ng L(-1)). The dynamics of target compounds were also investigated to provide a general trend of the level of parabens in a school indoor swimming pool during a 14-week period. Human exposure assessment was conducted to estimate the potential risk of exposure to parabens and their chlorinated derivatives in swimming pools. Considering the total exposure dose of multiple parabens, human exposure to parabens from the water of swimming pools is negligible. However, the threat of these parabens to children in swimming pool should be concerned.
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl 4-hydroxybenzoate, 99%
Sigma-Aldrich
6-Methylpurine, ≥99%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Heptyl 4-hydroxybenzoate, AldrichCPR
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99.0% (GC)
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Ethyl 4-hydroxybenzoate, ReagentPlus®, 99%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Nitrogen, ≥99.998%