Skip to Content
Merck
All Photos(1)

Key Documents

CDS001278

Sigma-Aldrich

Heptyl 4-hydroxybenzoate

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€85.60

€85.60


Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 G
€85.60

About This Item

Empirical Formula (Hill Notation):
C14H20O3
CAS Number:
Molecular Weight:
236.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

€85.60


Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

solid

SMILES string

CCCCCCCOC(=O)c1ccc(O)cc1

InChI

1S/C14H20O3/c1-2-3-4-5-6-11-17-14(16)12-7-9-13(15)10-8-12/h7-10,15H,2-6,11H2,1H3

InChI key

ZTJORNVITHUQJA-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuji Ogawa et al.
The Journal of toxicological sciences, 43(9), 557-563 (2018-09-07)
There is sometimes controversy over whether or not statistically significant responses produced in carcinogenicity studies have biologically significance. Ambiguous results from our previous two-year oral carcinogenicity study on acotiamide hydrochloride hydrate (acotiamide-HH), a prokinetic drug for functional dyspepsia, in rats
Wan-Li Ma et al.
Environment international, 95, 30-35 (2016-08-02)
Parabens are widely used as antimicrobial preservatives during pharmaceutical production. However, little information is available regarding the occurrence of parabens in commercial pharmaceuticals and their implications for human exposure. In this study, six commonly used parabens were analyzed by ultra-performance
Jef Stappaerts et al.
International journal of pharmaceutics, 529(1-2), 442-450 (2017-07-15)
Several studies have demonstrated the presence of aggregates in aqueous cyclodextrin containing solutions. The presence of guest compounds has been shown to influence this cyclodextrin aggregation process. In an attempt to gain insight into the effect of the physicochemical properties
Jinglan Feng et al.
Ecotoxicology and environmental safety, 172, 480-487 (2019-02-10)
In this study, six alkyl esters of p-hydroxybenzoic acids (parabens) and their metabolite, 4-hydroxybenzoic acid (p-HB) were simultaneously determined in surface water and sediment from the Yellow River and the Huai River in Henan Province, China. Concentrations of ∑parabens in
Rajendiran Karthikraj et al.
The Science of the total environment, 593-594, 592-598 (2017-04-01)
Parabens (p-hydroxy benzoic acid esters) are antimicrobial agents, used widely as preservatives in personal care products (PCPs), pharmaceuticals, foods, and beverages. PCPs that contain parabens are a major source of these chemicals in sewage treatment plants (STPs). Very few studies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service