Merck
  • Reduction of concentrated nitrate by using in situ synthesized zero-valent copper.
Reduction of concentrated nitrate by using in situ synthesized zero-valent copper.
Water science and technology : a journal of the International Association on Water Pollution Research (2015-09-12)
Tihitinna Asmellash Belay, F M Lin, C Y Lin, H M Hsiao, M F Chang, J C Liu
ABSTRACT
Although zero-valent iron represents a promising approach for reduction of nitrate (NO(3)(-)) in water, its application in concentrated nitrate is limited by surface passivation. In this study, an alternative approach using in situ synthesized zero-valent copper (Cu(0)) produced by borohydride (NaBH(4)) was investigated. Complete reduction was observed within 55 min by reacting 677 mg-N/L of NO(3)(-) with CuO (0.312 g/L) and NaBH(4) (4.16 g/L) at 60 °C. The pseudo-first-order rate constant was 0.059 min(-1), and it increased threefold when the CuO dose was increased to 1.24 g/L. Increasing the NaBH(4) dose produced less nitrite (NO(2)(-)) throughout the experiments, indicating that it is the primary agent for reducing NO(2)(-). The initial pH exerted a significant effect on the reaction rate, and NO(3)(-) was rapidly reduced when the initial pH was less than 4. Based on the research findings, possible reaction pathways for NO(3)(-) reduction by Cu(0) are proposed in this work.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%