Merck
  • Broad spectrum analysis of polar and apolar organic compounds in submicron atmospheric particles.
Broad spectrum analysis of polar and apolar organic compounds in submicron atmospheric particles.
Journal of chromatography. A (2015-06-10)
Marta Fontal, Barend L van Drooge, Jordi F López, Pilar Fernández, Joan O Grimalt
ABSTRACT
A method for the quantitative analysis of organic compounds on submicron particulate matter (PM1) collected on quartz filters was developed. The compounds analyzed encompassed C22-C35 alkanes, polycyclic aromatic hydrocarbons (PAHs), quinones, levoglucosan, cis-pinonic acid and short chain dicarboxylic acids such as malonic, succinic, glutaric, adipic, suberic, azelaic, malic and phthalic acids. The method included extraction with a pressure liquid extraction system, sample filtration though glass fibre filter, fractionation by high performance liquid chromatography and subsequent analysis by gas chromatography coupled to mass spectrometry. The study of the extraction efficiency of different solvent mixtures showed that DCM:MeOH 1:1 was the one providing the highest recoveries for all compounds. Extraction temperatures of 100°C provided better results than 60°C or 80°C. This method provided comparable extraction efficiency and qualitative and quantitative data to those involving Soxhlet extraction. Method recoveries for alkanes, most PAH, quinones and polar compounds calculated from spiked real samples were 52-72%, 78-101%, 50-62% and 76-104%, respectively, reproducibilities were 2-28%, 7-29%, 10-27% and 5-28%, respectively, limits of quantification were 0.01-0.1ng/m(3), 0.01-0.27ng/m(3), 0.04ng/m(3) and 0.32-2.8ng/m(3), respectively, which affords the quantification of a broad number of primary and secondary organic constituents of submicron aerosols.
Sigma-Aldrich
Fluorene, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Pyrene, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Benzo[ghi]perylene, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Phenyldodecane, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anthracene, reagent grade, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Benzo[k]fluoranthene, for fluorescence, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetone, natural, ≥97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Pyrene, sublimed grade, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Benzo[b]fluoranthene, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anthraquinone, 97%
SKU
Pack Size
Availability
Price
Quantity
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, NMR reference standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Succinic acid, ReagentPlus®, BioRenewable, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Succinic acid, BioXtra, BioRenewable, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Succinic acid, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Pyridine, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Benz[a]anthracene, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Suberic acid, 98%