Merck
  • Analysis of Hypodermic Needles and Syringes for the Presence of Blood and Polydimethylsiloxane (Silicone) Utilizing Microchemical Tests and Infrared Spectroscopy.
Analysis of Hypodermic Needles and Syringes for the Presence of Blood and Polydimethylsiloxane (Silicone) Utilizing Microchemical Tests and Infrared Spectroscopy.
Journal of forensic sciences (2015-03-19)
John B Crowe, Adam Lanzarotta, Mark R Witkowski, Sara E Andria
ABSTRACT
Suspect hypodermic needles and syringes were seized from an unlicensed individual who was allegedly injecting patients with silicone (polydimethylsiloxane [PDMS]) for cosmetic enhancement. Since control syringe barrels and needles often contain an interfering PDMS lubricant, a risk for false positives of foreign PDMS exists. The focus of this report was to minimize this risk and determine a quick and reliable test for the presence of blood in PDMS matrices. Using ATR-FT-IR spectroscopy, the risk for false-positive identification of foreign PDMS was reduced by (i) overfilling the sampling aperture to prevent spectral distortions and (ii) sampling a region of the suspect syringe/needle assembly where manufacturer-applied PDMS is not typically located. Analysis for blood indicated that the Teichman microchemical test was effective for detecting blood in the presence of PDMS. Overall, detecting PDMS established intent and detecting blood established that the needle containing the PDMS had been used for injection.
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Glucose solution, BioUltra, for molecular biology, ~20% in H2O
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
α-D-Glucose, anhydrous, 96%
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture