Merck
  • Chain length affects pancreatic lipase activity and the extent and pH-time profile of triglyceride lipolysis.
Chain length affects pancreatic lipase activity and the extent and pH-time profile of triglyceride lipolysis.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-05-06)
Paloma Benito-Gallo, Alessandro Franceschetto, Jonathan C M Wong, Maria Marlow, Vanessa Zann, Peter Scholes, Pavel Gershkovich
ABSTRACT
Triglycerides (TG) are one of the most common excipients used in oral lipid-based formulations. The chain length of the TG plays an important role in the oral bioavailability of the co-administered drug. Fatty acid (FA) chain-length specificity of porcine pancreatic lipase was studied by means of an in vitro lipolysis model under bio-relevant conditions at pH 6.80. In order to determine the total extent of lipolysis, back-titration experiments at pH 11.50 were performed. Results suggest that there is a specific chain length range (C2-C8) for which pancreatic lipase shows higher activity. This specificity could result from a combination of physicochemical properties of TGs, 2-monoglycerides (2-MGs) and FAs, namely the droplet size of the TGs, the solubility of 2-MGs within mixed micelles, and the relative stability of the FAs as leaving groups in the hydrolysis reaction. During experimentation, it was evident that an optimisation of lipolysis conditions was needed for tighter control over pH levels so as to better mimic in vivo conditions. 1M NaOH, 3.5 mL/min maximum dosing rate, and 3 μL/min minimum dosing rate were the optimised set of conditions that allowed better pH control, as well as the differentiation of the lipolysis of different lipid loads.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Glyceryl trioctanoate, ≥95% (GC)
Sigma-Aldrich
Glyceryl tridecanoate, ≥99% (GC)
Sigma-Aldrich
Glyceryl tributyrate, ≥99%
Sigma-Aldrich
Taurocholic acid sodium salt hydrate, BioXtra, ≥95% (HPLC)
Sigma-Aldrich
Taurocholic acid sodium salt hydrate, ≥95% (HPLC)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium taurocholate hydrate, ≥97.0% (TLC)
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O