Merck
  • Pharmacokinetic, bioavailability, metabolism and plasma protein binding evaluation of NADPH-oxidase inhibitor apocynin using LC-MS/MS.
Pharmacokinetic, bioavailability, metabolism and plasma protein binding evaluation of NADPH-oxidase inhibitor apocynin using LC-MS/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-02-16)
Hardik Chandasana, Yashpal S Chhonker, Veenu Bala, Yarra D Prasad, Telaprolu K Chaitanya, Vishnu L Sharma, Rabi S Bhatta
ABSTRACT
Apocynin is a major active constituent of Picrorhiza kurroa that exhibits potent anti-inflammatory activity by inhibiting superoxide-generating NADPH oxidase enzyme. To elucidate detailed pharmacokinetic profile of apocynin, high-performance liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS) method was developed in rat and human plasma. To the best of our knowledge, this is the first method for complete validation of apocynin in biological matrix using LC-MS/MS. Apocynin was rapidly absorbed after oral administration at 50mg/kg in rats and peak plasma level achieved within 5min. Moreover, plasma levels were observed up to 48h. The bioavailibity of apocynin was found to be 8.3%. In vitro plasma protein binding was found to be 83.41-86.07% and 71.39-73.34% in rat and human plasma, respectively. Apocynin was found stable in gastric (pH 1.2), intestinal (pH 6.8) and physiological (pH 7.4) fluids including microsomal (rat and human) stability studies. Further, apocynin did not convert to its dimeric form diapocynin in any of these studies. The data presented here provide crucial information about apocynin to support its pharmacological efficacy and further development as a potential anti-inflammatory drug candidate.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
USP
Phenacetin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
4′-Hydroxy-3′-methoxyacetophenone, 98%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Acetovanillone, ≥98%, FG
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material