Merck
  • Anionic nanoparticles based on Span 80 as low-cost, simple and efficient non-viral gene-transfection systems.
Anionic nanoparticles based on Span 80 as low-cost, simple and efficient non-viral gene-transfection systems.
International journal of pharmaceutics (2014-09-28)
A Pensado, I Fernandez-Piñeiro, B Seijo, A Sanchez
ABSTRACT
The existing strategies in the design of non-viral vectors for gene therapy are primarily conceived for cationic systems. However, the safety concerns associated with the use of positively charged systems for nucleic acid delivery and several reports regarding the efficacy of negatively charged systems highlights the need for improved gene-delivery vectors. With these premises in mind, we investigated the development of new negatively charged nanoparticles based on Sorbitan esters (Span(®)) – extremely cheap excipients broadly used in the pharmaceutical industry – on the basis of a simple, one-step and easily scalable procedure. For their specific use in gene therapy, we have incorporated oleylamine (OA) or poly-L-arginine (PA) into these nanosystems. Thus, we used Sorbitan monooleate (Span(®) 80) to design Span(®) 80-oleylamine and Span(®) 80-poly-L-arginine nanosystems (SP-OA and SP-PA, respectively). These systems can associate with the model plasmid pEGFP-C3 and show mean particle sizes of 304 nm and 247 nm and surface charges of -13 mV and -17 mV, respectively. The nanoparticles developed were evaluated in terms of in vitro cell viability and transfection ability. Both systems exhibited an appropriate cell-toxicity profile and are able to transfect the plasmid effectively. Specifically, the nanosystems including OA among their components provided higher transfection levels than the SP-PA nanoparticles. In conclusion, anionic nanoparticles based on Span(®) 80 can be considered low-cost, simple and efficient non-viral anionic gene-transfection systems.
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, absolute, over molecular sieve (H2O ≤0.04%), ≥99.5% (GC)
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, reagent grade
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, ≥99.0% (GC)
Supelco
Trehalose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Trehalose, United States Pharmacopeia (USP) Reference Standard
SAFC
Oleylamine, ≥98% (primary amine)
Supelco
Span® 80, suitable for GC
Supelco
Span® 80, viscosity 1000-2000 mPa.s (20 °C)
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder