Skip to Content
Merck
All Photos(1)

Key Documents

193453

Sigma-Aldrich

1,3-Dimethyl-2-imidazolidinone

reagent grade

Synonym(s):

N,N′-Dimethylethyleneurea, DMEU, DMI

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€28.10
100 G
€45.20
1 KG
€295.00

€28.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€28.10
100 G
€45.20
1 KG
€295.00

About This Item

Empirical Formula (Hill Notation):
C5H10N2O
CAS Number:
Molecular Weight:
114.15
Beilstein:
108808
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€28.10


Check Cart for Availability

Request a Bulk Order

grade

reagent grade

Quality Level

Assay

97.5 % (GC)

form

liquid

refractive index

n20/D 1.472 (lit.)

bp

224-226  °C
224-226 °C (lit.)

mp

46.8  °F

solubility

toluene: soluble(lit.)
water: miscible

density

1.056  g/mL at 25 °C
1.056 g/mL at 25 °C (lit.)

SMILES string

CN1CCN(C)C1=O
CN1CCN(C)C1=O

InChI

1S/C5H10N2O/c1-6-3-4-7(2)5(6)8/h3-4H2,1-2H3

InChI key

CYSGHNMQYZDMIA-UHFFFAOYSA-N
CYSGHNMQYZDMIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Dimethyl-2-imidazolidinone constitutes the mobile phase during size-exclusion chromatographic analysis of cellulose[1].

Application

1,3-Dimethyl-2-imidazolidinone was used as substitute solvent for toxic HMPA in the synthesis of 1,2-bis(trimethylsilyl)benzene[2]. It was employed as solvent during α-regioselective prenylation of imines[3].

Other Notes

Solvent used in various synthetic organic transformations. Studied in the formation of functionally stabilized hydrosilanediyl-transition metal complexes produced photochemically from arylsilanes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SEC-MALLS analysis of cellulose using LiCl/1, 3-dimethyl-2-imidazolidinone as an eluent.
Yanagisawa M, et al.
Cellulose, 11(2), 169-176 (2004)
Yolanda T Chong et al.
Cell, 161(6), 1413-1424 (2015-06-06)
Proteomics has proved invaluable in generating large-scale quantitative data; however, the development of systems approaches for examining the proteome in vivo has lagged behind. To evaluate protein abundance and localization on a proteome scale, we exploited the yeast GFP-fusion collection
K Krüger et al.
Scandinavian journal of medicine & science in sports, 25(3), e283-e291 (2014-09-30)
Different types of exercise are characterized by the ability to induce specific physiological stimuli that might be able to induce the mobilization of progenitor cells. The aim of the current study was to investigate the mobilization of hematopoietic progenitor cells
Li-Ming Zhao et al.
Organic letters, 14(3), 886-889 (2012-01-24)
A highly α-regioselective prenylation of imines has been successfully developed. The efficiency of this approach is confirmed by a wide range of imines including N- and C-aryl aldimines, N-alkyl aldimines, C-alkyl aldimines, and N- and C-aryl ketimines. The approach uses
Tsugio Kitamura et al.
The Journal of organic chemistry, 78(7), 3421-3424 (2013-03-20)
A practical and safe synthesis of 1,2-bis(trimethylsilyl)benzene from 1,2-dichlorobenzene and Me3SiCl was achieved by use of a hybrid metal of Mg and CuCl in the presence of LiCl in 1,3-dimethyl-2-imidazolidinone (DMI). This method does not require a toxic HMPA, provides

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service