Merck
  • Epidermal growth factor receptor-targeted immunoliposomes for delivery of celecoxib to cancer cells.
Epidermal growth factor receptor-targeted immunoliposomes for delivery of celecoxib to cancer cells.
International journal of pharmaceutics (2015-01-18)
Yanuar Dwi Putra Limasale, Ayşen Tezcaner, Can Özen, Dilek Keskin, Sreeparna Banerjee
ABSTRACT
Cyclooxygenase-2 (COX-2) is highly expressed in many different cancers. Therefore, the inhibition of the COX-2 pathway by a selective COX-2 inhibitor, celecoxib (CLX), may be an alternative strategy for cancer prevention and therapy. Liposomal drug delivery systems can be used to increase the therapeutic efficacy of CLX while minimizing its side effects. Previous studies have reported the encapsulation of CLX within the non-targeted long circulating liposomes and functional effect of these formulations against colorectal cancer cell lines. However, the selectivity and internalization of CLX-loaded liposomes can further be improved by grafting targeting ligands on their surface. Cetuximab (anti-epidermal growth factor receptor - EGFR - monoclonal antibody) is a promising targeting ligand since EGFR is highly expressed in a wide range of solid tumors. The aim of this study was to develop EGFR-targeted immunoliposomes for enhancing the delivery of CLX to cancer cells and to evaluate the functional effects of these liposomes in cancer cell lines. EGFR-targeted ILs, having an average size of 120nm, could encapsulate 40% of the CLX, while providing a sustained drug release profile. Cell association studies have also shown that the immunoliposome uptake was higher in EGFR-overexpressing cells compared to the non-targeted liposomes. In addition, the CLX-loaded-anti-EGFR immunoliposomes were significantly more toxic compared to the non-targeted ones in cancer cells with EGFR-overexpression but not in the cells with low EGFR expression, regardless of their COX-2 expression status. Thus, selective targeting of CLX with anti-EGFR immunoliposomes appears to be a promising strategy for therapy of tumors that overexpress EGFR.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Supelco
Rhodamine B, analytical standard
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Sigma-Aldrich
1,2-Distearoyl-sn-glycero-3-phosphocholine, ≥99%
Sigma-Aldrich
Rhodamine B, Dye content 90 %
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Rhodamine B, for fluorescence
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Supelco
Rhodamine B solution, 0.2% in isopropanol, for TLC derivatization
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%