Merck
  • Ultrasonic extraction with ultra-performance liquid chromatography/tandem mass spectrometry for the determination of ochratoxin A in processed cereal products.
Ultrasonic extraction with ultra-performance liquid chromatography/tandem mass spectrometry for the determination of ochratoxin A in processed cereal products.
Journal of AOAC International (2015-04-24)
Kieu Thi Ngoc Nguyen, Dojin Ryu
ABSTRACT
A rapid, simple, and reliable method using ultra-performance LC/MS/MS (UPLC/MS/MS) was developed for determination of ochratoxin A (OTA) in processed cereal products. OTA was ultrasonically extracted from the sample with acetonitrile-water (80 + 20, v/v), and the extract was then injected into the UPLC/MS/MS system after filtration. The calibration curves had good linearity with coefficients of determination greater than 0.999. Recoveries of OTA were in the range of 90-104%. LOD and LOQ of OTA in samples were 0.6 and 2.0 ng/g, respectively, and no significant matrix effect was found. This method was applied to determine OTA in 25 oat-based cereal samples. OTA was detected in five samples (20%) in the range of 2.4 to 7.3 ng/g.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ultrapure Acetonitrile
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%