Merck
  • Resistance of Neisseria meningitidis to human serum depends on T and B cell stimulating protein B.
Resistance of Neisseria meningitidis to human serum depends on T and B cell stimulating protein B.
Infection and immunity (2015-01-15)
Maike G Müller, Nina E Moe, Phillip Q Richards, Gregory R Moe
ABSTRACT
The ability of the human bacterial pathogen Neisseria meningitidis to cause invasive disease depends on survival in the bloodstream via mechanisms to suppress complement activation. In this study, we show that prophage genes coding for T and B cell stimulating protein B (TspB), which is an immunoglobulin-binding protein, are essential for survival of N. meningitidis group B strain H44/76 in normal human serum (NHS). H44/76 carries three genes coding for TspB. Mutants having all tspB genes inactivated did not survive in >5% NHS or IgG-depleted NHS. TspB appeared to inhibit IgM-mediated activation of the classical complement pathway, since survival of the tspB triple knockout was the same as that of the parent strain or a complemented mutant when the classical pathway was inactivated by depleting NHS of C1q and was increased in IgM-depleted NHS. A mutant solely carrying tspB gene nmbh4476_0681 was as resistant as the parent strain, while mutants carrying only nmbh4476_0598 or nmbh4476_1698 were killed in ≥5% NHS. The phenotype associated with TspB is formation of a matrix containing TspB, IgG, and DNA that envelopes aggregates of bacteria. Recombinant proteins corresponding to particular subdomains of TspB were found to have human IgG Fcγ- and/or DNA-binding activity, but only TspB derivatives containing both domains formed large, biofilm-like aggregates when combined with purified IgG and DNA. Recognizing the role of TspB in serum resistance may lead to a better understanding of why strains that carry tspB genes are associated with invasive meningococcal disease.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)