Merck
  • Discrimination of cathinone regioisomers, sold as 'legal highs', by Raman spectroscopy.
Discrimination of cathinone regioisomers, sold as 'legal highs', by Raman spectroscopy.
Drug testing and analysis (2013-08-01)
R Christie, E Horan, J Fox, C O'Donnell, H J Byrne, S McDermott, J Power, P Kavanagh
ABSTRACT
The discrimination of a cross section of cathinone regioisomers, sold as 'legal highs', using Raman spectroscopy, is reported here. Mephedrone and flephedrone were identified in 'legal high' products sold in Irish head shops, and their 2, 3 and 4-isomers were synthesized as reference standards. The 3,4-methylenedioxy substituted cathinones, methylone, butylone and methylenedioxypyrovalerone (MDPV), were also identified in 'legal highs' and their 2,3-isomers were synthesized for comparison. In addition, alpha- and beta-naphyrone were synthesized. Raman spectra of all the isomers were obtained using far-red excitation (785 nm) and it was found possible to discriminate the isomers of each substituted cathinone. In addition, Raman spectra were also recorded for a number of head shop products and, by comparison with the reference standards, correct isomer assignment for 4-mephedrone, 3-flephedrone, 3,4-methylone, 3,4-butylone, 3,4-MDPV, alpha-naphyrone and beta-naphyrone was achieved, thus providing a non-destructive, high-throughput and minimal sample preparation technique for the discrimination of such drug isomers.
Product Description
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Caffeine, European Pharmacopoeia (EP) Reference Standard
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Lidocaine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Lidocaine, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Lidocaine, United States Pharmacopeia (USP) Reference Standard