Merck
  • Surgical stress induced depressive and anxiety like behavior are improved by dapsone via modulating NADPH oxidase level.
Surgical stress induced depressive and anxiety like behavior are improved by dapsone via modulating NADPH oxidase level.
Neuroscience letters (2014-12-02)
Tao Zhang, Xiaosheng Tian, Qiudian Wang, Yawei Tong, Hecheng Wang, Zhengqian Li, Lunxu Li, Ting Zhou, Rui Zhan, Lei Zhao, Yang Sun, Dongsheng Fan, Lin Lu, Jing Zhang, Yinglan Jin, Weizhong Xiao, Xiangyang Guo, Dehua Chui
ABSTRACT
Surgical stress induced depression and anxiety like behavior are common complications among aged individuals suffering from surgery. Recent studies proposed that accumulation of oxidative stress is involved in the etiology of stress induced depression and anxiety. Dapsone possesses antioxidant properties, however, whether dapsone is effective in modulating surgical stress induced brain oxidative damage remains uncertain. The present study aimed to investigate the effect of dapsone on surgical stress induced depressive and anxiety like behavior, and brain oxidative stress in a well-established surgical stress model. Depressive and anxiety like behavior accompanied by elevated brain oxidative stress were observed in aged mice underwent abdominal surgery. Pretreatment with 5 mg/kg dapsone significantly improved the behavioral disorder and ameliorated brain oxidative stress in this model. Further investigation, revealed that surgical stress increased brain NADPH oxidase level, while pretreatment with dapsone abrogated the elevation of NADPH oxidase triggered by surgical stress. These findings suggest that dapsone is effective in improving surgical stress induced brain oxidative damage via down-regulating NADPH oxidase level in aged mice.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Dapsone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Supelco
Dapsone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dapsone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dapsone, VETRANAL®, analytical standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
4-Aminophenyl sulfone, 97%
Supelco
4,4′-Diaminodiphenylmethane, analytical standard
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)