Merck
Anticancer and anti-inflammatory activities of some dietary cucurbits.
Indian journal of experimental biology (2015-05-28)
Dhara Sharma, Indu Rawat, H C Goel
ABSTRACT
In this study, we investigated few dietary cucurbits for anticancer activity by monitoring cytotoxic (MTT and LDH assays), apoptotic (caspase-3 and annexin-V assays), and also their anti-inflammatory effects by IL-8 cytokine assay. Aqua-alcoholic (50:50) whole extracts of cucurbits [Lagenaria siceraria (Ls), Luffa cylindrica (Lc) and Cucurbita pepo (Cp)] were evaluated in colon cancer cells (HT-29 and HCT-15) and were compared with isolated biomolecule, cucurbitacin-B (Cbit-B). MTT and LDH assays revealed that the cucurbit extracts and Cbit-B, in a concentration dependent manner, decreased the viability of HT-29 and HCT-15 cells substantially. The viability of lymphocytes was, however, only marginally decreased, yielding a potential advantage over the tumor cells. Caspase-3 assay revealed maximum apoptosis with Ls while annexin V assay demonstrated maximum efficacy of Lc in this context. These cucurbits have also shown decreased secretion of IL-8, thereby revealing their anti-inflammatory capability. The results have demonstrated the therapeutic potential of dietary cucurbits in inhibiting cancer and inflammatory cytokine.
Product Description
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, PCR Reagent
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sign Into View Organizational & Contract Pricing
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sign Into View Organizational & Contract Pricing
Supelco
Water, for TOC analysis
Sign Into View Organizational & Contract Pricing
Supelco
Water, suitable for ion chromatography
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sign Into View Organizational & Contract Pricing
Supelco
Water, ACS reagent, for ultratrace analysis
Sign Into View Organizational & Contract Pricing
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sign Into View Organizational & Contract Pricing
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, BioPerformance Certified
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, ACS reagent
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, Deionized
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, suitable for HPLC
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Propidium iodide solution
Sign Into View Organizational & Contract Pricing