Merck
  • "Triple-punch" strategy for triple negative breast cancer therapy with minimized drug dosage and improved antitumor efficacy.
"Triple-punch" strategy for triple negative breast cancer therapy with minimized drug dosage and improved antitumor efficacy.
ACS nano (2015-01-23)
Shishuai Su, Yanhua Tian, Yiye Li, Yanping Ding, Tianjiao Ji, Meiyu Wu, Yan Wu, Guangjun Nie
ABSTRACT
Effective therapeutics against triple negative breast cancer (TNBC), which has no standard-of-care therapy, needs to be developed urgently. Here we demonstrated a strategy of integrating indocyanine green (ICG), paclitaxel (PTX), and survivin siRNA into one thermosensitive poly(2-(2-methoxyethoxy)ethyl methacrylate-co-oligo(ethylene glycol) methacrylate)-co-2-(dimethylamino)ethyl methacrylate-b-poly(D,L-lactide-co-glycolide) (P (MEO2MA-co-OEGMA-co-DMAEMA)-b-PLGA) nanoparticle (NP-IPS) for triple-punch strategy against TNBC. The NP-IPS significantly enhanced the stability of ICG. Controlled release of the PTX in tumor regions was triggered by the hyperthermia produced by laser irradiated ICG. The NP-IPS exhibited remarkable antitumor efficacy (almost complete ablation of the tumor xenografts) due to the combinational effects of chemotherapy, photothermal therapy, and gene therapy with low drug dose (ICG, 0.32 μmol/kg; PTX, 0.54 μmol/kg; siRNA, 1.5 mg/kg) and minimal side effects. Taken together, our current study demonstrates a nanoplatform for triple-therapy, which reveals a promising strategy for TNBC treatment.
Product Description
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Supelco
Pyrene, analytical standard
Supelco
Pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Luperox® A75, Benzoyl peroxide, 75%, remainder water
Sigma-Aldrich
Di(ethylene glycol) methyl ether methacrylate, 95%
Sigma-Aldrich
Luperox® A75FP, Benzoyl peroxide, 75% remainder water, contains 25 wt. % water as stabilizer, 75%
Sigma-Aldrich
Benzoyl peroxide blend with dicyclohexyl phthalate, suitable for use as a catalyst for electron microscopy. Modified to render it safe in transit.
Pyrene, BCR®, certified reference material
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Cell Counting Kit - 8, for quantitation of viable cell number in proliferation and cytotoxicity assays
Sigma-Aldrich
Cardiogreen, for microscopy
Sigma-Aldrich
Cardiogreen, polymethine dye
Sigma-Aldrich
Glycolide, ≥99%
Sigma-Aldrich
Ethylene glycol 5 M solution