Merck
  • Expression and purification of bioactive high-purity recombinant mouse SPP1 in Escherichia coli.
Expression and purification of bioactive high-purity recombinant mouse SPP1 in Escherichia coli.
Applied biochemistry and biotechnology (2014-03-26)
Yunsheng Yuan, Xiyuan Zhang, Shunyan Weng, Wen Guan, Di Xiang, Jin Gao, Jingjing Li, Wei Han, Yan Yu
ABSTRACT
Secreted phosphoprotein 1 (SPP1) is a phosphorylated acidic glycoprotein. It is broadly expressed in a variety of tissues, and it is involved in a number of physiological and pathological events, including cancer metastasis, tissues remodeling, pro-inflammation regulation, and cell survival. SPP1 has shown its function of protecting tissues and organs against injury and wound, giving itself potentials to become a therapy target or giving its antibodies of other counter-acting reagents potentials to become drug candidates. Non-tagged (native) recombinant SPP1 would be valuable in therapeutic and pharmaceutical researches. In our study, mouse Spp1 DNA fragment without signal peptide was built in pET28a(+) vector and transformed into Escherichia coli BL21 (DE3). The recombinant mouse SPP1 (rmSPP1) was then expressed in bacteria upon induction by isopropyl β-D-thiogalactopyranoside (IPTG). The abundance of rmSPP1 was increased using isoelectric precipitation and ammonium sulfate fractionation methods, and anion and cation exchange chromatography was employed to further purify rmSPP1. Finally, we got rmSPP1 product with 12.8 % productivity, 97 % purity, satisfactory bioactivity, and low endotoxin content.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sgpp1
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Spp1