Merck
  • Surface functionalized mesoporous silica nanoparticles as an effective carrier for epirubicin delivery to cancer cells.
Surface functionalized mesoporous silica nanoparticles as an effective carrier for epirubicin delivery to cancer cells.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-12-17)
Mohammad Yahya Hanafi-Bojd, Mahmoud Reza Jaafari, Navid Ramezanian, Min Xue, Mohamadreza Amin, Nasser Shahtahmassebi, Bizhan Malaekeh-Nikouei
ABSTRACT
Recent studies with inorganic nanoparticles modified with functional groups have demonstrated improvement in drug delivery into cancer cells. In the present study, we prepared, characterized, and evaluated mesoporous silica nanoparticles (MSNs) as carriers for epirubicin hydrochloride (EPI) in order to improve the antitumor efficacy of this drug. MSNs were prepared and functionalized with phosphonate, polyethylene glycol (PEG) and polyethylenimine-polyethylene glycol (PEI-PEG) groups. Different nanoparticulate formulations were loaded with EPI. The in vitro cytotoxicity and the in vivo antitumor efficacy of MSNs containing EPI were evaluated versus free EPI. The EPI release from nanoparticles was shown to be pH-dependent. The size of MSNs functionalized with polyethyleneimine-polyethylene glycol (MSN-PEI-PEG) was 123.8 ± 4.8 nm. This formulation showed the best antitumor effects at an EPI dose of 9 mg/kg in C-26 colon carcinoma model. The biodistribution results proved that MSN-PEI-PEG-EPI had a higher tumor accumulation compared to free EPI, 3h after drug administration. The results indicated that this formulation could be effective nanocarriers for anti-tumor therapies.
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, ≥95%
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, purum, ≥95.0% (NMR)
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Dimethyl ether, puriss., ≥99.9% (GC)
Sigma-Aldrich
Dimethyl ether, puriss., ≥99.9% (GC)
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt, ≥96.0% (HPCE), solid
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
Dimethyl ether, ≥99.9%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, BioReagent, ≥96.0% (HPCE)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
SAFC
Hexadecyltrimethylammonium bromide, USP/NF
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%