Skip to Content
Merck
All Photos(1)

Key Documents

743925

Sigma-Aldrich

Dimethyl ether

≥99.9%

Synonym(s):

Methyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

290 G
€1,030.00

€1,030.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
290 G
€1,030.00

About This Item

Linear Formula:
(CH3)2O
CAS Number:
Molecular Weight:
46.07
Beilstein:
1730743
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

€1,030.00


Check Cart for Availability

Request a Bulk Order

vapor density

1.62 (vs air)

Quality Level

vapor pressure

>760 mmHg ( 25 °C)

Assay

≥99.9%

form

gas

autoignition temp.

662 °F

expl. lim.

27 %

bp

−24.8 °C (lit.)

mp

−141 °C (lit.)

functional group

ether

SMILES string

COC

InChI

1S/C2H6O/c1-3-2/h1-2H3

InChI key

LCGLNKUTAGEVQW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

500ml high pressure aluminum cylinder equipped with a stainless steel valve according to DIN 477-1.

Other Notes

For 743925-290G-EU and 743925-290G packaging: 500 mL of aluminium cylinder equipped with brass valve.

Recommended products

For 743925-290G-EU and 743925-290G: outlet fitting: DIN 477-1 no. 1, Z742160 or Z742161 are the recommended regulators.

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

-41.8 °F - closed cup

Flash Point(C)

-41 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshiyuki Gambe et al.
Scientific reports, 5, 8869-8869 (2015-03-10)
The development of high energy-density lithium-ion secondary batteries as storage batteries in vehicles is attracting increasing attention. In this study, high-voltage bipolar stacked batteries with a quasi-solid-state electrolyte containing a Li-Glyme complex were prepared, and the performance of the device
Aniruddha Roy et al.
Molecular pharmaceutics, 11(8), 2592-2599 (2014-02-26)
Taxanes are a class of anticancer agents with a broad spectrum and have been widely used to treat a variety of cancer. However, its long-term use has been hampered by accumulating toxicity and development of drug resistance. The most extensively
Andrei Chekkoury et al.
Biomedical optics express, 6(9), 3134-3148 (2015-09-30)
Optical mesoscopy extends the capabilities of biological visualization beyond the limited penetration depth achieved by microscopy. However, imaging of opaque organisms or tissues larger than a few hundred micrometers requires invasive tissue sectioning or chemical treatment of the specimen for
Jian Wei He et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 32(9), 1523-1530 (2015-08-08)
In an investigation of deoxynivalenol (DON)-transformation products by Devosia mutans 17-2-E-8, the major product was identified as 3-epi-DON. This DON-transformation product was analysed by liquid chromatography and identified by congruent retention time and UV/Vis spectrum, as well as mass spectrometric
Zhigui Su et al.
Molecular pharmaceutics, 11(6), 1823-1834 (2014-05-02)
In this study, a dual-targeting drug delivery system based on bovine serum albumin nanoparticles (BSA-NPs) modified with both lactoferrin (Lf) and mPEG2000 loading doxorubicin (DOX) was designed, and its blood-brain barrier (BBB) penetration and brain glioma cells targeting properties were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service