Merck
  • ALCAT1 controls mitochondrial etiology of fatty liver diseases, linking defective mitophagy to steatosis.
ALCAT1 controls mitochondrial etiology of fatty liver diseases, linking defective mitophagy to steatosis.
Hepatology (Baltimore, Md.) (2014-09-10)
Li Wang, Xiaolei Liu, Jia Nie, Jun Zhang, Scot R Kimball, Hai Zhang, Weiping J Zhang, Leonard S Jefferson, Zeneng Cheng, Qiuhe Ji, Yuguang Shi
ABSTRACT
Defective autophagy is implicated in the pathogenesis of nonalcoholic fatty liver diseases (NAFLD) through poorly defined mechanisms. Cardiolipin is a mitochondrial phospholipid required for bioenergetics and mitophagy from yeast to mammals. Here we investigated a role for ALCAT1 in the development of NAFLD. ALCAT1 is a lysocardiolipin acyltransferase that catalyzes pathological cardiolipin remodeling in several aging-related diseases. We show that the onset of diet-induced NAFLD caused autophagic arrest in hepatocytes, leading to oxidative stress, mitochondrial dysfunction, and insulin resistance. In contrast, targeted deletion of ALCAT1 in mice prevented the onset of NAFLD. ALCAT1 deficiency also restored mitophagy, mitochondrial architecture, mitochondrial DNA (mtDNA) fidelity, and oxidative phosphorylation. In support of a causative role of the enzyme in a mitochondrial etiology of the disease, hepatic ALCAT1 expression was significantly up-regulated in mouse models of NAFLD. Forced expression of ALCAT1 in primary hepatocytes led to multiple defects that are highly reminiscent of NAFLD, including steatosis, defective autophagy, and mitochondrial dysfunction, linking pathological cardiolipin remodeling by ALCAT1 to the pathogenesis of NAFLD.
Product Description
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%