Merck
  • Inhibition of CD40-induced N-Ras activation reduces leishmania major infection.
Inhibition of CD40-induced N-Ras activation reduces leishmania major infection.
Journal of immunology (Baltimore, Md. : 1950) (2015-03-20)
Sushmita Chakraborty, Ankita Srivastava, Mukesh Kumar Jha, Arathi Nair, Surya Prakash Pandey, Neetu Srivastava, Sangeeta Kumari, Shailza Singh, Musti V Krishnasastry, Bhaskar Saha
ABSTRACT
Leishmania major is a parasite that resides and replicates in macrophages. We previously showed that the parasite enhanced CD40-induced Raf-MEK-ERK signaling but inhibited PI3K-MKK-p38MAPK signaling to proleishmanial effects. As Raf and PI3K have a Ras-binding domain but exert opposite effects on Leishmania infection, we examined whether Ras isoforms had differential roles in Leishmania infection. We observed that L. major enhanced N-Ras and H-Ras expression but inhibited K-Ras expression in macrophages. L. major infection enhanced N-Ras activity but inhibited H-Ras and K-Ras activity. TLR2 short hairpin RNA or anti-TLR2 or anti-lipophosphoglycan Abs reversed the L. major-altered N-Ras and K-Ras expressions. Pam3CSK4, a TLR2 ligand, enhanced N-Ras expression but reduced K-Ras expression, indicating TLR2-regulated Ras expression in L. major infection. Whereas N-Ras silencing reduced L. major infection, K-Ras and H-Ras silencing enhanced the infection both in macrophages in vitro and in C57BL/6 mice. BALB/c-derived macrophages transduced with lentivirally expressed N-Ras short hairpin RNA and pulsed with L. major-expressed MAPK10 enhanced MAPK10-specific Th1-type response. CD40-deficient mice primed with these macrophages had reduced L. major infection, accompanied by higher IFN-γ but less IL-4 production. As N-Ras is activated by Sos, a guanine nucleotide exchange factor, we modeled the N-Ras-Sos interaction and designed two peptides from their interface. Both the cell-permeable peptides reduced L. major infection in BALB/c mice but not in CD40-deficient mice. These data reveal the L. major-enhanced CD40-induced N-Ras activation as a novel immune evasion strategy and the potential for Ras isoform-targeted antileishmanial immunotherapy and immunoprophylaxis.
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human KRAS
SAFC
Sodium chloride solution, 5 M
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
MISSION® esiRNA, targeting human NRAS
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%