Merck
  • Microfluidic conceived drug loaded Janus particles in side-by-side capillaries device.
Microfluidic conceived drug loaded Janus particles in side-by-side capillaries device.
International journal of pharmaceutics (2014-06-24)
Ikram Ullah Khan, Christophe A Serra, Nicolas Anton, Xiang Li, Roman Akasov, Nadia Messaddeq, Isabelle Kraus, Thierry F Vandamme
ABSTRACT
A side-by-side capillaries microfluidic device was developed to fabricate drug loaded poly(acrylamide)/poly(methyl acrylate) Janus particles in the range of 59-240 μm by UV-assisted free radical polymerization. This system was characterized in terms of continuous and dispersed phases flow rates (Qc/Qd), monomer composition of the two compartments, surfactant nature and concentration, outlet tube diameter and UV intensity. These factors were adequately controlled to get different particle shapes ranging from core-shell to bi-compartmental particles. For the latter, a low surfactant concentration (0.75 wt.%) was necessary when the two dispersed phases were pumped at equal flow rate, while at high surfactant concentration, dispersed phases flow rates have to be changed. FTIR analysis suggested complete polymerization of monomers and cytotoxicity test showed these particles were biocompatible having LD 50 of 9 mg/mL. Both ketoprofen and sodium fluorescein were released in sustained release manner at pH 6.8 by following a diffusion type release mechanism. Drug release was faster for bigger particles and found to result from the irregular distribution of the two phases and indentation on bigger particles as revealed by SEM analysis. In comparison, sodium fluorescein release was slower which was attributed to low encapsulation but could be modified by decreasing crosslinker concentration.
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Supelco
Methyl acrylate, analytical standard
Sigma-Aldrich
Methyl acrylate, 99%, contains ≤100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
1-Hydroxycyclohexyl phenyl ketone, 99%
Sigma-Aldrich
Ethyl acrylate, contains 10-20 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, for molecular biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Supelco
Acrylamide, analytical standard
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Supelco
Ethyl acrylate, analytical standard
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Ethyl acrylate, ≥99.5%, stabilized
Supelco
Fluorescein sodium salt, analytical standard
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Fluorescein sodium salt, BioReagent, suitable for fluorescence
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Supelco
Acrylamide solution, 40% in H2O, for molecular biology
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
N,N′-Methylenebisacrylamide solution, suitable for electrophoresis, 2% in H2O
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Fluorescein sodium salt, used as fluorescent tracer
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material