Merck
Kisspeptin modulates fertilization capacity of mouse spermatozoa.
Reproduction (Cambridge, England) (2014-02-26)
Meng-Chieh Hsu, Jyun-Yuan Wang, Yue-Jia Lee, De-Shien Jong, Kuan-Hao Tsui, Chih-Hsien Chiu
ABSTRACT
Kisspeptin acts as an upstream regulator of the hypothalamus-pituitary-gonad axis, which is one of the main regulatory systems for mammalian reproduction. Kiss1 and its receptor Kiss1r (also known as G protein-coupled receptor 54 (Gpr54)) are expressed in various organs, but their functions are not well understood. The purpose of this study was to investigate the expression profiles and functions of kisspeptin and KISS1R in the reproductive tissues of imprinting control region mice. To identify the expression pattern and location of kisspeptin and KISS1R in gonads, testes and ovarian tissues were examined by immunohistochemical or immunofluorescent staining. Kisspeptin and KISS1R were expressed primarily in Leydig cells and seminiferous tubules respectively. KISS1R was specifically localized in the acrosomal region of spermatids and mature spermatozoa. Kisspeptin, but not KISS1R, was expressed in the cumulus-oocyte complex and oviductal epithelium of ovarian and oviductal tissues. The sperm intracellular calcium concentrations significantly increased in response to treatment with kisspeptin 10 in Fluo-4-loaded sperm. The IVF rates decreased after treatment of sperm with the kisspeptin antagonist peptide 234. These results suggest that kisspeptin and KISS1R might be involved in the fertilization process in the female reproductive tract. In summary, this study indicates that kisspeptin and KISS1R are expressed in female and male gametes, respectively, and in mouse reproductive tissues. These data strongly suggest that the kisspeptin system could regulate mammalian fertilization and reproduction.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)