Merck
  • Acquired resistance to gemcitabine and cross-resistance in human pancreatic cancer clones.
Acquired resistance to gemcitabine and cross-resistance in human pancreatic cancer clones.
Anti-cancer drugs (2014-09-05)
Hiroshi Yoneyama, Asako Takizawa-Hashimoto, Osamu Takeuchi, Yukiko Watanabe, Koichiro Atsuda, Fumiki Asanuma, Yoshinori Yamada, Yukio Suzuki
ABSTRACT
The efficacy of gemcitabine (GEM), a standard treatment agent for pancreatic cancer, is insufficient because of primary or acquired resistance to this drug. Patients with tumors intrinsically sensitive to GEM gradually acquire resistance and require a shift to second agents, which are associated with the risk of cross-resistance. However, whether cross-resistance is actually present has long been disputed. Using six GEM-resistant and four highly GEM-resistant clones derived from the pancreatic cancer cell line BxPC-3, we determined the resistance of each clone and parent cell line to GEM and four anticancer agents (5-FU, CDDP, CPT-11, and DTX). The GEM-resistant clones had different resistances to GEM and other agents, and did not develop a specific pattern of cross-resistance. This result shows that tumor cells are heterogeneous. However, all highly GEM-resistant clones presented overexpression of ribonucleotide reductase subunit M1 (RRM1), a target enzyme for metabolized GEM, and showed cross-resistance with 5-FU. The expression level of RRM1 was high; therefore, resistance to GEM was high. We showed that a tumor cell acquired resistance to GEM, and cross-resistance developed in one clone. These results suggest that only cells with certain mechanisms for high-level resistance to GEM survive against selective pressure applied by highly concentrated GEM. RRM1 may be one of the few factors that can induce high resistance to GEM and a suitable therapeutic target for GEM-resistant pancreatic cancer.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Irinotecan hydrochloride, topoisomerase inhibitor
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium succinate dibasic hexahydrate, puriss. p.a., ≥98.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
5-Fluorouracil, analytical standard
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium succinate dibasic hexahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium succinate dibasic hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.