Merck
  • Comparison of liquid chromatography and supercritical fluid chromatography coupled to compact single quadrupole mass spectrometer for targeted in vitro metabolism assay.
Comparison of liquid chromatography and supercritical fluid chromatography coupled to compact single quadrupole mass spectrometer for targeted in vitro metabolism assay.
Journal of chromatography. A (2014-12-03)
Dany Spaggiari, Florence Mehl, Vincent Desfontaine, Alexandre Grand-Guillaume Perrenoud, Szabolcs Fekete, Serge Rudaz, Davy Guillarme
ABSTRACT
The goal of this study was to evaluate the combination of powerful chromatographic methods and compact single quadrupole MS device for simple in vitro cytochrome P450 (CYP) inhibition assay, instead of more expensive triple quadrupole MS/MS detectors. For this purpose, two modern chromatographic approaches (ultra-high pressure liquid chromatography (UHPLC) and ultra-high performance supercritical fluid chromatography (UHPSFC)) were tested in combination with simple MS detector. To show the applicability for an in vitro CYP-mediated metabolism assay using the cocktail approach, a method was first developed in UHPLC-MS to separate a mixture of 8 probe substrates and 8 CYP-specific metabolites. A screening procedure was initially applied to determine the best combination of a column, an organic modifier and a mobile-phase pH, followed by fine tuning of the conditions (i.e., gradient profile, temperature and pH) using HPLC modelling software. A similar sequential method development procedure was also evaluated for UHPSFC-MS. For method development, where peak tracking is necessary, the use of single quadrupole MS was found to be extremely valuable for following the investigated analytes. Ultimately, a baseline separation of the 16 compounds was achieved in both UHPLC-MS and UHPSFC-MS with an analysis time of less than 7 min. In a second series of experiments, sensitivity was evaluated, and LOQ values were between 2 and 100 ng/mL in UHPLC-MS, while they ranged from 2 to 200 ng/mL in UHPSFC-MS. Based on the concentrations employed for the current in vitro phase I metabolism assay, these LOQ values were appropriate for this type of application. Finally, the two analytical methods were applied to in vitro CYP-dependent metabolism testing. Two well-known phytochemical inhibitors, yohimbine and resveratrol, were investigated, and reliable conclusions were drawn from these experiments with both UHPLC-MS and UHPSFC-MS. At the end, the proposed strategy of optimized chromatography combined with simple MS device has been shown to potentially compete with the widely used combination of generic chromatography and highly selective MS/MS device for simple in vitro CYP inhibition assays. In addition, our analytical method may be easier to use in a routine environment; the instrument cost is significantly reduced and the two developed methods fit for purpose.
Product Description
Coumarin, primary reference standard
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Phenacetin, Pharmaceutical Secondary Standard; Certified Reference Material
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
6-Hydroxychlorzoxazone, ≥98% (HPLC)
Supelco
Coumarin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
SAFC
HEPES
Supelco
Heptane, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Coumarin, European Pharmacopoeia (EP) Reference Standard
SAFC
HEPES
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Acetic acid, analytical standard
Supelco
Methanol, analytical standard
Supelco
Heptane, analytical standard
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
Sigma-Aldrich
Bupropion hydrochloride, ≥98% (HPLC), solid
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Yohimbine hydrochloride, ≥98% (HPLC), powder