Merck
  • Optimization of extraction parameters on the isolation of phenolic compounds from sour cherry (Prunus cerasus L.) pomace.
Optimization of extraction parameters on the isolation of phenolic compounds from sour cherry (Prunus cerasus L.) pomace.
Journal of food science and technology (2015-04-22)
Fatih Mehmet Yılmaz, Mehmet Karaaslan, Hasan Vardin
ABSTRACT
There is an increasing interest to bio-components obtained from fruit & vegetable wastes. Anthocyanin is one of the components regained from fruits especially from red ones. Sour cherry is considered as industrial fruit since most of the grown fruit is processed into juice and hence considerable amount of pomace is removed from process. The influences of process parameters on the extraction of phenolic compounds from sour cherry pomace were investigated. Fifty-one percent ethanol concentration, 75 °C temperature and 12 mL/g solvent to solid ratio were selected as optimum process parameters. Time effect on the process efficiency was monitored at three different temperatures (25, 50 and 75 °C) and total phenolic and total anthocyanin contents were found to reach equilibrium concentrations between 80 and 100 min time intervals. Ethanol concentrations affected differently extraction yield of anthocyanins and non-anthocyanin phenolics. Various individual phenolic compounds present in sour cherry pomace were identified and quantified by HPLC equipped with UV/Vis PDA. Of the phenolic compounds, cyanidin-3-glucosylrutinoside, neochlorogenic acid and catechin were the most abundant ones found in the pomace. 14.23 ± 0.38 mg/g total phenolic, 0.41 ± 0.02 mg/g total anthocyanin, 0.19 ± 0.02 mg/g cyanidin-3-glucosylrutinoside, 0.22 ± 0.01 mg/g neochlorogenic acid and 0.22 ± 0.02 mg/g catechin contents (dry weight) were determined in the pomace at optimum extraction conditions.
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Neochlorogenic acid, from Lonicera japonica, ≥98.0% (HPLC)
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)