Skip to Content
Merck

Skip To

94419

Neochlorogenic acid

from Lonicera japonica, ≥98.0% (HPLC)

Synonym(s):

5-O-(trans-3,4-Dihydroxycinnamoyl)-D-quinic acid, trans-5-O-Caffeoylquinic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
€ 186,00
50 mg
Check Cart for Availability
€ 726,00

About This Item

Empirical Formula (Hill Notation):
C16H18O9
CAS Number:
Molecular Weight:
354.31
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
212-997-1
MDL number:

€ 186,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Lonicera japonica

Quality Segment

assay

≥98.0% (HPLC)

form

powder or crystals

loss

≤3.0% loss on drying

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

O[C@@H]1C[C@@](O)(C[C@@H](OC(=O)\C=C\c2ccc(O)c(O)c2)[C@H]1O)C(O)=O

InChI

1S/C16H18O9/c17-9-3-1-8(5-10(9)18)2-4-13(20)25-12-7-16(24,15(22)23)6-11(19)14(12)21/h1-5,11-12,14,17-19,21,24H,6-7H2,(H,22,23)/b4-2+/t11-,12-,14+,16-/m1/s1

InChI key

CWVRJTMFETXNAD-NXLLHMKUSA-N

General description

Neochlorogenic acid is a polyphenolic compound and an ester of caffeic acid and quinic acid.[1][2] It is an isomer of Chlorogenic acid.[2][3]

Application

Neochlorogenic acid has been used:
  • to examine the phenolic content in the roots and aerial parts of Serratula centauroides using high performance liquid chromatography (HPLC) technique
  • to study the anti-inflammatory response of dried plum in human synovial fibroblasts (hSF) and bone resorption in rheumatoid arthritic transgenic mice[1]
  • for the quantitative and toxicity analysis of green coffee bean extract on human liver carcinoma cell line (HepG2 cells)[2]

Biochem/physiol Actions

Neochlorogenic acid has been found to have an inhibitory effect on the biosynthesis of melanin. Therefore it can be used on harvested fruits to prevent brown rot infection caused by Monilinia laxa.[3] It also shows impressive antioxidant, antibacterial, antiviral and antipyretic characteristics.[4] Neochlorogenic acid has the potential to be used in the management of acute and chronic inflammatory diseases due to its anti-inflammatory activity.[5] It may function as chemopreventive compounds due to growth inhibition seen in breast cancer cell line MDA-MB-435.[6]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHL80504C3878SMB00131
form

powder or crystals

form

-

form

powder

form

solid

biological source

Lonicera japonica

biological source

-

biological source

-

biological source

-

assay

≥98.0% (HPLC)

assay

≥95.0% (HPLC)

assay

≥95% (titration)

assay

≥95% (LC/MS-ELSD)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

application(s)

food and beverages

application(s)

-

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

Quality Level

100

Quality Level

-

Quality Level

200

Quality Level

200

loss

≤3.0% loss on drying

loss

-

loss

-

loss

-


Still not finding the right product?

Explore all of our products under Neochlorogenic acid


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters