Merck
  • In vitro inhibition of lysine decarboxylase activity by organophosphate esters.
In vitro inhibition of lysine decarboxylase activity by organophosphate esters.
Biochemical pharmacology (2014-09-30)
Sufang Wang, Bin Wan, Lianying Zhang, Yu Yang, Liang-Hong Guo
ABSTRACT
Organophosphate esters (OPEs), a major group of organophosphorus flame retardants, are regarded as emerging environmental contaminants of health concern. Amino acid decarboxylases catalyze the conversion of amino acids into polyamines that are essential for cell proliferation, hypertrophy and tissue growth. In this paper, inhibitory effect of twelve OPEs with aromatic, alkyl or chlorinated alkyl substituents on the activity of lysine decarboxylase (LDC) was assessed quantitatively with an economic and label-free fluorescence sensor and cell assay. The sensor comprises a macrocyclic host (cucurbit[7]uril) and a fluorescent dye (acridine orange) reporter. The twelve OPEs were found to vary in their capacity to inhibit LDC activity. Alkyl group substituted OPEs had no inhibitory effect. By contrast, six OPEs substituted with aromatic or chlorinated alkyl groups inhibited LDC activity significantly with IC50 ranging from 1.32 μM to 9.07 μM. Among them, the inhibitory effect of tri-m-cresyl phosphate (TCrP) was even more effective as an inhibitor than guanosine 5'-diphosphate-3'-diphosphate (ppGpp) (1.60 μM), an LDC natural inhibitor in vivo. Moreover, at non-cytotoxic concentrations, these six OPEs showed perceptible inhibitory effects on LDC activity in PC12 living cells, and led to a marked loss in the cadaverine content. Molecular docking analysis of the LDC/OPE complexes revealed that different binding modes contribute to the difference in their inhibitory effect. Our finding suggested that LDC, as a new potential biological target of OPEs, might be implicated in toxicological and pathogenic mechanism of OPEs.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Cadaverine, 95%
Sigma-Aldrich
1,7-Diaminoheptane, 98%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Benzoyl chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Cadaverine, purum, ≥97.0% (GC)
Sigma-Aldrich
Benzoyl chloride, 99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Supelco
Cadaverine, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis