Merck
  • Investigations on the transfer of porphyrin from o/w emulsion droplets to liposomes with two different methods.
Investigations on the transfer of porphyrin from o/w emulsion droplets to liposomes with two different methods.
Drug development and industrial pharmacy (2013-10-30)
Mohamed Dawoud
ABSTRACT
Due to their small particle size, colloidal fat emulsions are suitable for intravenous administration. In order to obtain information on their potential in vivo performance, it is important to find a simple and effective in vitro assay to evaluate the drug release behavior of such particles. Two in vitro methods were studied to measure the transfer of a lipophilic model drug from colloidal o/w emulsion droplets (donor) to liposomes (acceptor), which serve as model membranes mimicking cell membranes in the body. In the first method (column method) the acceptor particles were neutral unilamellar vesicles. In the second method (MLV method), multilamellar vesicles (MLV) were used as acceptor. The donor nanoemulsions were prepared by high pressure homogenization. Z-average particle size, polydispersity index and zeta potential were determined. The transfer of porphyrin was moderate to the acceptor MLV and rapid to the acceptor unilamellar vesicles. The amount of transferred porphyrin at the end of the experiment depended on the transfer method and the donor/acceptor ratio. With both acceptors the transfer of porphyrin stopped at a concentration lower than the theoretical equilibrium values. Many factors such as acceptor particle size and donor to acceptor lipid molar ratio affect the drug transfer from the donor particles to the different acceptors. Both methods seem to be suitable to study the drug transfer from such colloidal emulsion and the use of lipophilic acceptor particles is a better approach to the conditions in blood.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
HEPES
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%