Merck
  • High performance liquid chromatography tandem mass spectrometry dual extraction method for identification of green tea catechin metabolites excreted in human urine.
High performance liquid chromatography tandem mass spectrometry dual extraction method for identification of green tea catechin metabolites excreted in human urine.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-10-13)
Kayleigh A Clarke, Tristan P Dew, Rachel E B Watson, Mark D Farrar, Susan Bennett, Anna Nicolaou, Lesley E Rhodes, Gary Williamson
ABSTRACT
The simultaneous analysis of free-form and conjugated flavonoids in the same sample is difficult but necessary to properly estimate their bioavailability. A method was developed to optimise the extraction of both free and conjugated forms of catechins and metabolites in a biological sample following the consumption of green tea. A double-blind randomised controlled trial was performed in which 26 volunteers consumed daily green tea and vitamin C supplements and 24 consumed a placebo for 3 months. Urine was collected for 24h at 4 separate time points (pre- and post-consumption) to confirm compliance to the supplementation and to distinguish between placebo and supplementation consumption. The urine was assessed for both free and conjugated metabolites of green tea using LC-MS(2) analysis, after a combination extraction method, which involved an ethyl acetate extraction followed by an acetonitrile protein precipitation. The combination method resulted in a good recovery of EC-O-sulphate (91±7%), EGC-O-glucuronide (94±6%), EC (95±6%), EGC (111±5%) and ethyl gallate (74±3%). A potential total of 55 catechin metabolites were investigated, and of these, 26 conjugated (with methyl, glucuronide or sulphate groups) and 3 free-form (unconjugated) compounds were identified in urine following green tea consumption. The majority of EC and EGC conjugates significantly increased post-consumption of green tea in comparison to baseline (pre-supplementation) samples. The conjugated metabolites associated with the highest peak areas were O-methyl-EC-O-sulphate and the valerolactones M6/M6'-O-sulphate. In line with previous studies, EC and EGC were only identified as conjugated derivatives, and EGCG and ECG were not found as mono-conjugated or free-forms. In summary, the method reported here provides a good recovery of catechin compounds and is appropriate for use in the assessment of flavonoid bioavailability, particularly for biological tissues that may contain endogenous deconjugating enzymes.
Product Description
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Syringic acid, ≥95% (HPLC)
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
3-Hydroxybenzoic acid, ReagentPlus®, 99%
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
USP
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Benzoic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Syringic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Hippuric acid, 98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline