Merck
  • TCAB1: a potential target for diagnosis and therapy of head and neck carcinomas.
TCAB1: a potential target for diagnosis and therapy of head and neck carcinomas.
Molecular cancer (2014-07-30)
Chong-kui Sun, Xiao-bo Luo, Ya-ping Gou, Ling Hu, Kun Wang, Chao Li, Zhen-ting Xiang, Ping Zhang, Xiang-li Kong, Chao-liang Zhang, Qin Yang, Jing Li, Li-ying Xiao, Yan Li, Qian-ming Chen
ABSTRACT
WRAP53, including α, β and γ isoforms, plays an important role not only in the stability of p53 mRNA, but also in the assembly and trafficking of the telomerase holoenzyme. It has been considered an oncogene and is thought to promote the survival of cancer cells. The aim of this study was to detect the role of TCAB1 (except WRAP53α) in the occurrence and development of head and neck carcinomas. Immunohistochemistry was used to detect the TCAB1 expression in clinical specimen sections and performed western blotting to check the TCAB1 expression levels in cell lines. TCAB1 was depleted using shRNA lentivirus and the knockdown efficiency was assessed using q-PCR and Western blotting. We performed CCK-8 assays and flow cytometry to check the cell proliferation potential and used the trans-well assay to test the invasion ability in vitro. Xenografts were used to detect the tumor formation potential in vivo. Moreover, we performed cDNA microarray to investigate the candidate factors involved in this process. We observed a notable overexpression of TCAB1 in head and neck carcinoma clinical specimens as well as in carcinoma cell lines. Knockdown of TCAB1 decreased the cellular proliferation potential and invasion ability in vitro. cDNA microarray analysis suggested the possible involvement of several pathways and factors associated with tumorigenesis and carcinoma development in the TCAB1-mediated regulation of cancers. Furthermore, the xenograft assay confirmed that the depletion of TCAB1 would inhibit tumor formation in nude mice. The immunohistochemistry results of the mice tumor tissue sections revealed that the cells in shTCAB1 xenografts showed decreased proliferation potential and increased apoptotic trend, meanwhile, the angiogenesis was inhibited in the smaller tumors form shTCAB1 cells. Our study demonstrated that depletion of TCAB1 decreased cellular proliferation and invasion potential both in vitro and in vivo. The data indicated that TCAB1 might facilitate the occurrence and development of head and neck carcinomas. In future, TCAB1 might be useful as a prognostic biomarker or a potential target for the diagnosis and therapy of head and neck carcinomas.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Clca2
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Anti-WRAP53 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
MISSION® esiRNA, targeting human CLCA2
Sigma-Aldrich
MISSION® esiRNA, targeting human CLCA1
Sigma-Aldrich
DAPI, for nucleic acid staining