Merck
  • F-actin flow drives affinity maturation and spatial organization of LFA-1 at the immunological synapse.
F-actin flow drives affinity maturation and spatial organization of LFA-1 at the immunological synapse.
The Journal of cell biology (2015-02-11)
William A Comrie, Alexander Babich, Janis K Burkhardt
ABSTRACT
Integrin-dependent interactions between T cells and antigen-presenting cells are vital for proper T cell activation, effector function, and memory. Regulation of integrin function occurs via conformational change, which modulates ligand affinity, and receptor clustering, which modulates valency. Here, we show that conformational intermediates of leukocyte functional antigen 1 (LFA-1) form a concentric array at the immunological synapse. Using an inhibitor cocktail to arrest F-actin dynamics, we show that organization of this array depends on F-actin flow and ligand mobility. Furthermore, F-actin flow is critical for maintaining the high affinity conformation of LFA-1, for increasing valency by recruiting LFA-1 to the immunological synapse, and ultimately for promoting intracellular cell adhesion molecule 1 (ICAM-1) binding. Finally, we show that F-actin forces are opposed by immobilized ICAM-1, which triggers LFA-1 activation through a combination of induced fit and tension-based mechanisms. Our data provide direct support for a model in which the T cell actin network generates mechanical forces that regulate LFA-1 activity at the immunological synapse.
Product Description
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, suitable for ion chromatography
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Nitrilotriacetic acid trisodium salt, Sigma Grade, ≥98%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Anti-ITGB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC