Merck
  • Sesquiterpene lactones of Moquiniastrum polymorphum subsp. floccosum have antineoplastic effects in Walker-256 tumor-bearing rats.
Sesquiterpene lactones of Moquiniastrum polymorphum subsp. floccosum have antineoplastic effects in Walker-256 tumor-bearing rats.
Chemico-biological interactions (2015-01-24)
Gracianny Gomes Martins, Francislaine Aparecida dos Reis Lívero, Aline Maria Stolf, Caroline Machado Kopruszinski, Cibele Campos Cardoso, Olair Carlos Beltrame, José Ederaldo Queiroz-Telles, Regiane Lauriano Batista Strapasson, Maria Élida Alves Stefanello, Ronald Oude-Elferink, Alexandra Acco
ABSTRACT
This study aimed to evaluate the in vivo antitumor actions and toxicity of the dichloromethane fraction (F1B) of Moquiniastrum polymorphum subsp. floccosum (formerly Gochnatia polymorpha ssp. floccosa), composed of sesquiterpene lactones, against Walker-256 carcinosarcoma in rats. Male Wistar rats received 100 mg kg(-1) F1B per day orally for 16 days after subcutaneous inoculation of Walker-256 cells in the pelvic limb. The tumor progression was monitored, and after treatment, tumor weight, oxidative stress, plasma biochemistry, inflammatory parameters, gene expression and histology of tumor and/or liver were evaluated. The toxicity of F1B was analyzed through the relative weight of organs. Additionally, an LD50 test was performed in mice. F1B treatment significantly reduced tumor volume and weight. There was no difference in oxidative stress in tumor tissue after treatment. F1B treatment modified hepatic glutathione and superoxide dismutase, and normalized plasma glucose, alkaline phosphatase, and amylase. F1B did not affect the activity of myeloperoxidase and N-acetylglucosaminidase or the nitric oxide levels in tumor tissue. However, F1B decreased the tumor necrosis factor (TNF)-α levels. Additionally, F1B increased apoptosis in the tumor, mediated by up-regulation of the p53 and Bax gene expression. No clinical signs of toxicity or death were observed in the rats treated with F1B. The LD50 calculated for mice was 1209 mg kg(-1). F1B, which is rich in sesquiterpene lactones, showed antitumor activity against Walker-256 carcinosarcoma. This effect may be, at least in part, related to the induction of apoptosis and inhibition of TNF-α signaling.
Product Description
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Supelco
Sodium nitrite, analytical standard
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent