Merck
  • Establishment of opioid-induced rewarding effects under oxaliplatin- and Paclitaxel-induced neuropathy in rats.
Establishment of opioid-induced rewarding effects under oxaliplatin- and Paclitaxel-induced neuropathy in rats.
Journal of pharmacological sciences (2014-08-22)
Tomohisa Mori, Tomoe Kanbara, Masato Harumiya, Yoshiyuki Iwase, Aki Masumoto, Sachiko Komiya, Atsushi Nakamura, Masahiro Shibasaki, Toshiyuki Kanemasa, Gaku Sakaguchi, Tsutomu Suzuki
ABSTRACT
The rewarding effects of μ-receptor agonists can be suppressed under several pain conditions. We recently showed that clinically used μ-receptor agonists possess efficacies for relieving the neuropathic pain induced by chemotherapeutic drug in rats; however, it is possible that the use of μ-receptor agonists may trigger the rewarding effects even under chemotherapeutic drug-induced neuropathic pain. Nevertheless, no information is available regarding whether μ-receptor agonists produce psychological dependence under chemotherapeutic drug-induced neuropathic pain. Therefore, we examined the effects of neuropathy induced by chemotherapeutic drugs on the rewarding effects of morphine, oxycodone, and fentanyl in rats. Repeated treatment with oxaliplatin or paclitaxel produced neuropathy as measured by the von Frey test. Rewarding effects produced by antinociceptive doses of μ-receptor agonists were not suppressed under oxaliplatin- or paclitaxel-induced neuropathy. Furthermore, the morphine-induced increase in the release of dopamine from the nucleus accumbens, which is a critical step in the rewarding effects of μ-receptor agonists, was not altered in paclitaxel-treated rats. These results suggest that the rewarding effects of μ-receptor agonists can still be established under oxaliplatin- or paclitaxel-induced neuropathic pain. Therefore, patients should be carefully monitored for psychological dependence on μ-receptor agonists when they are used to control chemotherapeutic drug-induced neuropathic pain.
Sigma-Aldrich
α-D-Glucose, anhydrous, 96%
Sigma-Aldrich
Glucose solution, BioUltra, for molecular biology, ~20% in H2O
Supelco
Oxaliplatin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
Oxaliplatin, powder
Oxaliplatin, European Pharmacopoeia (EP) Reference Standard
USP
Oxaliplatin, United States Pharmacopeia (USP) Reference Standard