Skip to Content
Merck
All Photos(1)

Key Documents

PHR1528

Supelco

Oxaliplatin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Oxaliplatin, [SP-4-2-(1R-trans)]-(1,2-Cyclohexanediamine-N,N′)[ethanedioata(2--)-O,O’]platinum

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
€361.00

€361.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 MG
€361.00

About This Item

Empirical Formula (Hill Notation):
C8H14N2O4Pt
CAS Number:
Molecular Weight:
397.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€361.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000271
traceable to USP 1481204

API family

oxaliplatin

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

O=C1O[Pt]OC1=O.N[C@@H]2CCCC[C@H]2N

InChI

1S/C6H14N2.C2H2O4.Pt/c7-5-3-1-2-4-6(5)8;3-1(4)2(5)6;/h5-6H,1-4,7-8H2;(H,3,4)(H,5,6);/q;;+2/p-2/t5-,6-;;/m1../s1

InChI key

ZROHGHOFXNOHSO-BNTLRKBRSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Oxaliplatin, a third generation anti-cancer drug platinum compound, is utilized for the treatment of colorectal cancer. It exhibits a better safety profile than cisplatin (first generation) and carboplatin (second generation).[1]

Application

Oxaliplatin may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography[2][3] and electrophoresis[1][4] techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB8974 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Lact. - Muta. 2 - Repr. 1B - Resp. Sens. 1B - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of a liposomal oxaliplatin drug formulation by capillary electrophoresis hyphenated to inductively coupled plasma mass spectrometry (CE-ICP-MS).
Nguyen TTTN, et al.
Analytical and Bioanalytical Chemistry, 402(6), 2131-2139 (2012)
Quality control of pharmaceutical formulations containing cisplatin, carboplatin, and oxaliplatin by micellar and microemulsion electrokinetic chromatography (MEKC, MEEKC).
Nussbaumer S, et al.
Journal of Pharmaceutical and Biomedical Analysis, 55(2), 253-258 (2011)
Validation of a LC method for the analysis of oxaliplatin in a pharmaceutical formulation using an experimental design.
Ficarra R, et al.
Journal of Pharmaceutical and Biomedical Analysis, 29(6), 1097-1103 (2002)
Characterization of a liposome-based formulation of oxaliplatin using capillary electrophoresis: encapsulation and leakage.
Franzen U, et al.
Journal of Pharmaceutical and Biomedical Analysis, 55(1), 16-22 (2011)
L B J van Iersel et al.
European journal of surgical oncology : the journal of the European Society of Surgical Oncology and the British Association of Surgical Oncology, 40(11), 1557-1563 (2014-08-16)
To improve isolated hepatic perfusion (IHP), we performed a phase I dose-escalation study to determine the optimal oxaliplatin dose in combination with a fixed melphalan dose. Between June 2007 and July 2008, 11 patients, comprising of 8 colorectal cancer and

Questions

  1. What is the solubility of PHR1528 in saline (0.9% NaCl)

    1 answer
    1. This product is not tested for solubility. However, the research grade product of the identical compound, O9512, is tested for solubility in water at 5 mg/mL. A similar solubility rate would be expected in normal saline. See the link below to review a sample Certificate of Analysis:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/O9512-BULKMKCR8366.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service