Merck
  • Apolipoprotein E isoforms disrupt long-chain fatty acid distribution in the plasma, the liver and the adipose tissue of mice.
Apolipoprotein E isoforms disrupt long-chain fatty acid distribution in the plasma, the liver and the adipose tissue of mice.
Prostaglandins, leukotrienes, and essential fatty acids (2014-10-11)
Valérie Conway, Annie Larouche, Wael Alata, Milène Vandal, Frédéric Calon, Mélanie Plourde
ABSTRACT
Evidences suggest that omega-3 fatty acid (n-3 PUFA) metabolism is imbalanced in apolipoprotein E epsilon 4 isoform carriers (APOE4). This study aimed to investigate APOE genotype-dependant modulation of FA profiles, protein and enzyme important to fatty acid (FA) metabolism in the adipose tissue, the liver and the plasma using human APOE-targeted replacement mouse-model (N=37). FA transport (FATP) and binding (FABP) protein levels in tissues and concentrations of liver carnitine palmitoyltransferase 1 (CPT1) were performed. N-3 PUFA concentration was >45% lower in the adipose tissue and liver of APOE4 mice compared to APOE3 mice. In APOE4 mice, there were higher levels of FATP and FABP in the liver and higher FATP in the adipose tissue compared to APOE2 mice. There was a trend towards higher CPT1 concentrations in APOE4 mice compared to APOE3 mice. Therefore, since APOE-isoform differences were not always in line with the unbalanced n-3 PUFA profiles in organs, other proteins may be involved in maintaining n-3 PUFA homeostasis in mice with different APOE-isoforms.
Sigma-Aldrich
Boron, ≥95% (boron), amorphous powder
Boron, monofilament, 5m, diameter 0.1mm
Boron, monofilament, 5m, diameter 0.2mm
Boron, monofilament, 20m, diameter 0.2mm
Boron, monofilament, 20m, diameter 0.1mm
Boron, monofilament, 50m, diameter 0.2mm
Boron, monofilament, 10m, diameter 0.1mm
Boron, monofilament, 200m, diameter 0.1mm
Boron, monofilament, 10m, diameter 0.2mm
Boron, monofilament, 100m, diameter 0.1mm
Boron, monofilament, 50m, diameter 0.1mm
Sigma-Aldrich
Boron, crystalline, 1 cm, 99.7% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)