Skip to Content
Merck
All Photos(2)

Key Documents

GF97861247

Boron

monofilament, 20m, diameter 0.1mm

Synonym(s):

Boron, B 005910

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€307.00

€307.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
€307.00

About This Item

Empirical Formula (Hill Notation):
B
CAS Number:
Molecular Weight:
10.81
MDL number:
UNSPSC Code:
12141907
PubChem Substance ID:
NACRES:
NA.23

€307.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

monofilament

manufacturer/tradename

Goodfellow 978-612-47

resistivity

1.5E12 μΩ-cm, 20 °C

density

2.34 g/mL at 25 °C (lit.)

SMILES string

[B]

InChI

1S/B

InChI key

ZOXJGFHDIHLPTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne-Marie Alexander et al.
Chemical Society reviews, 39(11), 4388-4401 (2010-06-08)
Catalysts generated by the addition of carbon, nitrogen or phosphorus to transition metals have interesting properties and potential applications. The addition of carbon, nitrogen or phosphorus can lead to substantial modification of the catalytic efficacy of the parent metal and
Li Song et al.
Advanced materials (Deerfield Beach, Fla.), 24(36), 4878-4895 (2012-07-14)
Two-dimensional (2D) atomic layers derived from bulk layered materials are very interesting from both scientific and application viewpoints, as evidenced from the story of graphene. Atomic layers of several such materials such as hexagonal boron nitride (h-BN) and dichalcogenides are
Kyoko Miwa et al.
Advances in experimental medicine and biology, 679, 83-96 (2010-07-30)
Understanding of the molecular mechanisms of boron (B) transport has been greatly advanced in the last decade. BOR1, the first B transporter in living systems, was identified by forward genetics using Arabidopsis mutants. Genes similar to BOR1 have been reported
Ting Jiang et al.
Organic letters, 16(7), 1952-1955 (2014-03-29)
A novel stepwise and regioselective nucleophilic aromatic substitution (SNAr) reaction of halogenated boron dipyrrins (BODIPYs) with pyrroles has been developed under mild conditions with no catalyst needed and shown to be diversity oriented. The resultant pyrrole-substituted BODIPYs are interesting red
Monika A Wimmer et al.
Plant science : an international journal of experimental plant biology, 203-204, 25-32 (2013-02-19)
Boron (B) is an essential microelement for plants and is constantly needed throughout the plant life due to its function as a structural element of the plant cell wall. B deficiency is a wide-spread problem in agricultural areas world-wide, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service