Merck
  • Inulin-based polymer coated SPIONs as potential drug delivery systems for targeted cancer therapy.
Inulin-based polymer coated SPIONs as potential drug delivery systems for targeted cancer therapy.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-10-05)
C Scialabba, M Licciardi, N Mauro, F Rocco, M Ceruti, G Giammona
ABSTRACT
This paper deal with the synthesis and characterization of PEGylated squalene-grafted-inulin amphiphile capable of self-assembling and self-organizing into nanocarriers once placed in aqueous media. It was exploited as coating agent for obtaining doxorubicin loaded superparamagnetic iron oxide nanoparticles (SPIONs) endowed with stealth like behavior and excellent physicochemical stability. Inulin was firstly modified in the side chain with primary amine groups, followed in turn by conjugation with squalenoyl derivatives through common amidic coupling agents and PEGylation by imine linkage. Polymer coated SPIONs were so obtained by spontaneous self-assembling of inulin copolymer onto magnetite surface involving hydrophobic-hydrophobic interactions between the metallic core and the squalene moieties. The system was characterized in terms of hydrodynamic radius, zeta potential, shape and drug loading capacity. On the whole, the stealth-like shell stabilized the suspension in aqueous media, though allowing the release of the doxorubicin loaded in therapeutic range. The cytotoxicity profile on cancer (HCT116) cell line and in vitro drug uptake were evaluated both with and without an external magnetic field used as targeting agent and uptake promoter, displaying that magnetic targeting implies advantageous therapeutic effects, that is amplified drug uptake and increased anticancer activity throughout the tumor mass.
Product Description
Supelco
Squalene, analytical standard
Supelco
Ethylenediamine, analytical standard
Sigma-Aldrich
Ethylenediamine solution, technical, 75-80%
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
Ethylenediamine, puriss. p.a., absolute, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Bis(4-nitrophenyl) carbonate, ≥99%
Sigma-Aldrich
Squalene, ≥98%, liquid
Sigma-Aldrich
Ethylenediamine, meets USP testing specifications
Sigma-Aldrich
Ethylenediamine, BioXtra
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – 2-propanol solution, derivatization grade (GC derivatization), ~1.25 M HCl (T), LiChropur
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
N,N-Dimethylformamide, ≥99%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, BioReagent, 36.5-38.0%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Neocuproine, ≥98%
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Triethylamine, ≥99%
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur