Merck
  • Influence of postharvest processing and storage conditions on key antioxidants in pūhā (Sonchus oleraceus L.).
Influence of postharvest processing and storage conditions on key antioxidants in pūhā (Sonchus oleraceus L.).
The Journal of pharmacy and pharmacology (2014-04-05)
Zong-Quan Ou, David M Schmierer, Clare J Strachan, Thomas Rades, Arlene McDowell
ABSTRACT
To investigate effects of different postharvest drying processes and storage conditions on key antioxidants in Sonchus oleraceus L. leaves. Fresh leaves were oven-dried (60°C), freeze-dried or air-dried (∼25°C) for 6 h, 24 h and 3 days, respectively. Design of experiments (DOE) was applied to study the stability of antioxidants (caftaric, chlorogenic and chicoric acids) in S. oleraceus leaves and leaf extracts stored at different temperatures (4, 25 and 50°C) and relative humidities (15%, 43% and 75%) for 180 days. The concentration of antioxidants was quantified by a HPLC-2,2'-diphenylpicrylhydrazyl post-column derivatisation method. Antioxidant activity was assessed by a cellular antioxidant activity assay. The three antioxidants degraded to unquantifiable levels after oven-drying. More than 90% of the antioxidants were retained by freeze-drying and air-drying. Both leaf and extract samples retained >90% of antioxidants, except those stored at 75% relative humidity. Leaf material had higher antioxidant concentrations and greater cellular antioxidant activity than corresponding extract samples. Freeze-drying and air-drying preserved more antioxidants in S. oleraceus than oven-drying. From DOE analysis, humidity plays an important role in degradation of antioxidants during storage. To preserve antioxidant activity, it is preferable to store S. oleraceus as dried leaf material.
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Methanol, analytical standard
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%