Merck
  • Intestinal expression of Fas and Fas ligand is upregulated by bacterial signaling through TLR4 and TLR5, with activation of Fas modulating intestinal TLR-mediated inflammation.
Intestinal expression of Fas and Fas ligand is upregulated by bacterial signaling through TLR4 and TLR5, with activation of Fas modulating intestinal TLR-mediated inflammation.
Journal of immunology (Baltimore, Md. : 1950) (2014-11-08)
Philana Fernandes, Charlotte O'Donnell, Caitriona Lyons, Jonathan Keane, Tim Regan, Stephen O'Brien, Padraic Fallon, Elizabeth Brint, Aileen Houston
ABSTRACT
TLRs play an important role in mediating intestinal inflammation and homeostasis. Fas is best studied in terms of its function in apoptosis, but recent studies demonstrate that Fas signaling may mediate additional functions such as inflammation. The role of Fas, and the Fas ligand (FasL), in the intestine is poorly understood. The aim of this study was to evaluate potential cross-talk between TLRs and Fas/FasL system in intestinal epithelial cells (IECs). IECs were stimulated with TLR ligands, and expression of Fas and FasL was investigated. Treatment with TLR4 and TLR5 ligands, but not TLR2 and 9 ligands, increased expression of Fas and FasL in IECs in vitro. Consistent with this finding, expression of intestinal Fas and FasL was reduced in vivo in the epithelium of TLR4 knockout (KO), 5KO, and germ-free mice, but not in TLR2KO mice. Modulating Fas signaling using agonistic anti-Fas augmented TLR4- and TLR5-mediated TNF-α and IL-8 production by IECs. In addition, suppression of Fas in IECs reduced the ability of TLR4 and TLR5 ligands and the intestinal pathogens Salmonella typhimurium and Listeria monocytogenes to induce the expression of IL-8. In conclusion, this study demonstrates that extensive cross-talk in IECs occurs between the Fas and TLR signaling pathways, with the FasL/Fas system playing a role in TLR-mediated inflammatory responses in the intestine.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Resorufin, 95%
Sigma-Aldrich
TPCA-1, ≥95% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Anti-Fas Antibody (human, activating), clone CH11, clone CH11, Upstate®, from mouse