Merck
  • WNT Agonist Decreases Tissue Damage and Improves Renal Function After Ischemia-Reperfusion.
WNT Agonist Decreases Tissue Damage and Improves Renal Function After Ischemia-Reperfusion.
Shock (Augusta, Ga.) (2014-12-17)
Michael Kuncewitch, Weng-Lang Yang, Lana Corbo, Adam Khader, Jeffrey Nicastro, Gene F Coppa, Ping Wang
ABSTRACT
Renal ischemia-reperfusion (IR) injury (IRI) after shock states or transplantation causes tissue damage and delayed graft function, respectively. The Wnt/β-catenin signaling pathway plays a critical role in nephrogenesis. We therefore hypothesized that pharmacological activation of the Wnt/β-catenin signaling by the Wnt agonist, a synthetic pyrimidine, could protect kidneys from IRI. Adult male rats were subjected to bilateral clamping of the renal pedicles with microvascular clips for 60 min, followed by reperfusion. The Wnt agonist (5 mg/kg body weight) or vehicle (20% dimethyl sulfoxide in saline) was administered intravenously 1 h before ischemia. Blood and renal tissues were collected 24 h after IR for evaluation. Renal IR caused a significant reduction of β-catenin and its downstream target gene cyclin D1 by 65% and 39%, respectively, compared with the sham, whereas the Wnt agonist restored them to sham levels. The number and intensity of cells staining with the proliferation marker Ki67 in ischematized kidneys were enhanced by the Wnt agonist. The integrity of the renal histological architecture in the Wnt agonist group was better preserved than the vehicle group. The Wnt agonist significantly lowered serum levels of creatinine, aspartate aminotransferase, and lactate dehydrogenase and inhibited the production of interleukin 6 and interleukin 1β and myeloperoxidase activities. Lastly, the Wnt agonist reduced inducible nitric oxide synthase, nitrotyrosine proteins, and 4-hydroxynonenal in the kidneys by 60%, 47%, and 21%, respectively, compared with the vehicle. These results indicate that the Wnt agonist improves renal regeneration and function while attenuating inflammation and oxidative stress in the kidneys after IR. Thus, pharmacologic stimulation of the Wnt/β-catenin signaling provides a beneficial effect on the prevention of renal IRI.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Sodium chloride solution, 0.85%