Merck
  • A study of lipid- and water-soluble arsenic species in liver of Northeast Arctic cod (Gadus morhua) containing high levels of total arsenic.
A study of lipid- and water-soluble arsenic species in liver of Northeast Arctic cod (Gadus morhua) containing high levels of total arsenic.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS) (2015-01-27)
Veronika Sele, Jens J Sloth, Kåre Julshamn, Kasper Skov, Heidi Amlund
ABSTRACT
In the present study liver samples (n=26) of Northeast Arctic cod (Gadus morhua), ranging in total arsenic concentrations from 2.1 to 240mg/kg liver wet weight (ww), were analysed for their content of total arsenic and arsenic species in the lipid-soluble and water-soluble fractions. The arsenic concentrations in the lipid fractions ranged from 1.8 to 16.4mg As/kg oil of liver, and a linear correlation (r(2)=0.80, p<0.001) was observed between the total arsenic concentrations in liver and the total arsenic concentrations in the respective lipid fractions of the same livers. The relative proportion of arsenolipids was considerably lower in liver samples with high total arsenic levels (33-240mg/kg ww), which contained from 3 to 7% of the total arsenic in the lipid-soluble fraction. In contrast liver samples with low arsenic concentrations (2.1-33mg/kg ww) contained up to 50% of the total arsenic as lipid-soluble species. Arsenic speciation analysis of the lipid-soluble fractions of the livers, using reversed-phase high performance liquid chromatography coupled to inductively coupled plasma mass spectrometry (HPLC-ICP-MS), revealed the presence of several arsenolipids. Three major arsenic-containing hydrocarbons (C17H39AsO, C19H41AsO and C23H37AsO) and five arsenic-containing fatty acids (C17H35AsO3, C19H39AO3, C19H37AsO3, C23H37AsO3 and C24H37AsO3) were identified using HPLC coupled to quadrupole time-of-flight mass spectrometry (qTOF-MS). Arsenobetaine was the major arsenic species in the water-soluble fraction of the livers, while dimethylarsinate, arsenocholine and inorganic arsenic were minor constituents. Inorganic arsenic accounted for less than 0.1% of the total arsenic in the liver samples.
Sigma-Aldrich
Yttrium, chips, 99.9% trace rare earth metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Supelco
Methanol, analytical standard
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Thallium, granular, ≤6 mm diameter, 99.9% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Cesium, ingot, ≥99.95% trace metals basis
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Heptane, anhydrous, 99%
Supelco
Heptane, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Heptane, suitable for HPLC, ≥99%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Heptane, suitable for HPLC, ≥96%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ammonium carbonate, ACS reagent, ≥30.0% NH3 basis
Sigma-Aldrich
Methanol, Absolute - Acetone free
Yttrium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Yttrium, foil, 25mm disks, thickness 0.50mm, as rolled, 99%
Yttrium, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
Yttrium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%