Merck
  • Protective role of adenylate cyclase in the context of a live pertussis vaccine candidate.
Protective role of adenylate cyclase in the context of a live pertussis vaccine candidate.
Microbes and infection (2013-10-22)
Annabelle Lim, Jowin K W Ng, Camille Locht, Sylvie Alonso
ABSTRACT
Despite high vaccination coverage, pertussis remains an important respiratory infectious disease and the least-controlled vaccine-preventable infectious disease in children. Natural infection with Bordetella pertussis is known to induce strong and long-lasting immunity that wanes later than vaccine-mediated immunity. Therefore, a live attenuated B. pertussis vaccine, named BPZE1, has been developed and has recently completed a phase I clinical trial in adult human volunteers. In this study, we investigated the contribution of adenylate cyclase (CyaA) in BPZE1-mediated protection against pertussis. A CyaA-deficient BPZE1 mutant was thus constructed. Absence of CyaA did not compromise the adherence properties of the bacteria onto mammalian cells. However, the CyaA-deficient mutant displayed a slight impairment in the ability to survive within macrophages compared to the parental BPZE1 strain. In vivo, whereas the protective efficacy of the CyaA-deficient mutant was comparable to the parental strain at a vaccine dose of 5 × 10(5) colony forming units (CFU), it was significantly impaired at a vaccine dose of 5 × 10(3) CFU. This impairment correlated with impaired lung colonization ability, and impaired IFN-γ production in the animal immunized with the CyaA-deficient BPZE1 mutant while the pertussis-specific antibody profile and Th17 response were comparable to those observed in BPZE1-immunized mice. Our findings thus support a role of CyaA in BPZE1-mediated protection through induction of cellular mediated immunity.
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Supelco
Digoxigenin, analytical standard
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
HEPES, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C